Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153725263-153725492 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCD1
Entrez Gene ID
215
Gene Name
ATP binding cassette subfamily D member 1
Gene Aliases
ABC42, ALD, ALDP, AMN
UniGene
Hs.159546
Gene Chromosome Location
Chr.X: 153724851-153744762 on Build GRCh38
Transcripts
NM_000033.3

SNP Detail

SNP ID Chromosome Location
rs74315278 Chr.X: 153725333-153725349 on Build GRCh38
rs398123109 Chr.X: 153725297-153725312 on Build GRCh38
rs965462099 Chr.X: 153725321-153725321 on Build GRCh38
rs782480731 Chr.X: 153725325-153725325 on Build GRCh38
rs781828916 Chr.X: 153725411-153725411 on Build GRCh38
rs781989555 Chr.X: 153725444-153725444 on Build GRCh38
rs782454198 Chr.X: 153725358-153725358 on Build GRCh38
rs368718078 Chr.X: 153725374-153725374 on Build GRCh38
rs375019683 Chr.X: 153725366-153725366 on Build GRCh38
rs782611595 Chr.X: 153725328-153725328 on Build GRCh38
rs183021839 Chr.X: 153725304-153725304 on Build GRCh38
rs782122122 Chr.X: 153725298-153725298 on Build GRCh38
rs781837108 Chr.X: 153725455-153725455 on Build GRCh38
rs782397391 Chr.X: 153725413-153725413 on Build GRCh38
rs782693577 Chr.X: 153725316-153725316 on Build GRCh38
rs781900720 Chr.X: 153725306-153725306 on Build GRCh38
rs782161942 Chr.X: 153725307-153725307 on Build GRCh38
rs782447609 Chr.X: 153725464-153725464 on Build GRCh38
rs1057114018 Chr.X: 153725465-153725465 on Build GRCh38
rs782134465 Chr.X: 153725447-153725447 on Build GRCh38
rs782061571 Chr.X: 153725280-153725280 on Build GRCh38
rs782728047 Chr.X: 153725452-153725452 on Build GRCh38
rs781803154 Chr.X: 153725409-153725409 on Build GRCh38
rs782195097 Chr.X: 153725356-153725356 on Build GRCh38
rs781855598 Chr.X: 153725320-153725320 on Build GRCh38
rs782819655 Chr.X: 153725440-153725440 on Build GRCh38
rs782208191 Chr.X: 153725428-153725428 on Build GRCh38
rs782317790 Chr.X: 153725437-153725437 on Build GRCh38
rs782079729 Chr.X: 153725427-153725427 on Build GRCh38

View Full Product Details