Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 74394236-74394507 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADK
Entrez Gene ID
132
Gene Name
adenosine kinase
Gene Aliases
AK
UniGene
Hs.656586
Gene Chromosome Location
Chr.10: 74151185-74709303 on Build GRCh38
Transcripts
NM_001123.3 NM_001202449.1 NM_001202450.1 NM_006721.3 XM_017015698.1 XM_017015699.1 XM_017015700.1 XM_017015701.1 XM_017015702.1 XM_017015703.1 XM_017015704.1 XM_017015705.1 XM_017015706.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3440574 c.432C>T p.I144I Chr.10: 74394299-74394299 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs879369993 Chr.10: 74394372-74394372 on Build GRCh38
rs770870702 Chr.10: 74394443-74394443 on Build GRCh38
rs1042292179 Chr.10: 74394277-74394277 on Build GRCh38
rs142477111 Chr.10: 74394296-74394296 on Build GRCh38
rs1046361049 Chr.10: 74394474-74394474 on Build GRCh38
rs894592030 Chr.10: 74394432-74394432 on Build GRCh38
rs746933712 Chr.10: 74394301-74394301 on Build GRCh38
rs148712301 Chr.10: 74394293-74394293 on Build GRCh38
rs758588957 Chr.10: 74394303-74394303 on Build GRCh38
rs573088827 Chr.10: 74394335-74394335 on Build GRCh38
rs911230622 Chr.10: 74394423-74394423 on Build GRCh38
rs769097993 Chr.10: 74394331-74394331 on Build GRCh38
rs752472596 Chr.10: 74394276-74394276 on Build GRCh38
rs773486641 Chr.10: 74394351-74394351 on Build GRCh38
rs770168687 Chr.10: 74394345-74394345 on Build GRCh38
rs747117198 Chr.10: 74394330-74394330 on Build GRCh38
rs536543659 Chr.10: 74394273-74394273 on Build GRCh38
rs886047236 Chr.10: 74394308-74394308 on Build GRCh38
rs780071590 Chr.10: 74394310-74394310 on Build GRCh38
rs575724005 Chr.10: 74394460-74394460 on Build GRCh38
rs750638068 Chr.10: 74394302-74394302 on Build GRCh38
rs748657523 Chr.10: 74394340-74394340 on Build GRCh38

View Full Product Details