Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 58208914-58209187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTR10
Entrez Gene ID
55860
Gene Name
actin-related protein 10 homolog
Gene Aliases
ACTR11, Arp10, Arp11, HARP11
UniGene
Hs.509451
Gene Chromosome Location
Chr.14: 58200115-58235636 on Build GRCh38
Transcripts
NM_018477.2 XM_011536960.1 XM_011536961.1 XM_011536962.2 XM_011536963.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3496551 c.283G>A p.V95I Chr.14: 58209048-58209048 on Build GRCh38
6593066 c.234-7delT p.? Chr.14: 58208992-58208992 on Build GRCh38
6314187 c.290G>T p.C97F Chr.14: 58209055-58209055 on Build GRCh38
7174844 c.319C>T p.R107C Chr.14: 58209084-58209084 on Build GRCh38
2245911 c.282G>A p.S94S Chr.14: 58209047-58209047 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776707478 Chr.14: 58209098-58209098 on Build GRCh38
rs768258106 Chr.14: 58208949-58208949 on Build GRCh38
rs760788489 Chr.14: 58209017-58209017 on Build GRCh38
rs768075462 Chr.14: 58209006-58209006 on Build GRCh38
rs917303658 Chr.14: 58209112-58209112 on Build GRCh38
rs754031809 Chr.14: 58209025-58209025 on Build GRCh38
rs758308190 Chr.14: 58209118-58209118 on Build GRCh38
rs747767626 Chr.14: 58209050-58209050 on Build GRCh38
rs541821610 Chr.14: 58208994-58208994 on Build GRCh38
rs750299910 Chr.14: 58209029-58209029 on Build GRCh38
rs758255014 Chr.14: 58209035-58209035 on Build GRCh38
rs761124038 Chr.14: 58209085-58209085 on Build GRCh38
rs978731557 Chr.14: 58209004-58209004 on Build GRCh38
rs372194628 Chr.14: 58209080-58209080 on Build GRCh38
rs761598148 Chr.14: 58209119-58209119 on Build GRCh38
rs780189321 Chr.14: 58209039-58209039 on Build GRCh38
rs778579321 Chr.14: 58209028-58209028 on Build GRCh38
rs759962263 Chr.14: 58209001-58209001 on Build GRCh38
rs1013543883 Chr.14: 58208988-58208988 on Build GRCh38
rs772732171 Chr.14: 58209061-58209061 on Build GRCh38
rs557017236 Chr.14: 58209047-58209047 on Build GRCh38
rs113113758 Chr.14: 58208975-58208975 on Build GRCh38
rs768311278 Chr.14: 58209046-58209046 on Build GRCh38
rs751563170 Chr.14: 58209126-58209126 on Build GRCh38
rs753215827 Chr.14: 58208981-58208981 on Build GRCh38
rs774717336 Chr.14: 58208999-58208999 on Build GRCh38
rs866737585 Chr.14: 58209115-58209115 on Build GRCh38
rs376837714 Chr.14: 58209088-58209088 on Build GRCh38
rs754903055 Chr.14: 58209139-58209139 on Build GRCh38
rs868147529 Chr.14: 58209093-58209093 on Build GRCh38
rs769419229 Chr.14: 58209054-58209054 on Build GRCh38
rs777432690 Chr.14: 58209106-58209106 on Build GRCh38
rs1000390299 Chr.14: 58209081-58209081 on Build GRCh38
rs575489047 Chr.14: 58209111-58209111 on Build GRCh38
rs369023311 Chr.14: 58209114-58209114 on Build GRCh38
rs750588197 Chr.14: 58209117-58209117 on Build GRCh38
rs753185350 Chr.14: 58209011-58209011 on Build GRCh38
rs764122724 Chr.14: 58209024-58209024 on Build GRCh38
rs761484321 Chr.14: 58208991-58208991 on Build GRCh38
rs993238597 Chr.14: 58209077-58209077 on Build GRCh38
rs780569862 Chr.14: 58209145-58209145 on Build GRCh38
rs200649605 Chr.14: 58209042-58209042 on Build GRCh38
rs757444319 Chr.14: 58209027-58209027 on Build GRCh38
rs968588135 Chr.14: 58208973-58208973 on Build GRCh38
rs772399098 Chr.14: 58209084-58209084 on Build GRCh38

View Full Product Details