Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 49630720-49630971 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL11
Entrez Gene ID
115761
Gene Name
ADP ribosylation factor like GTPase 11
Gene Aliases
ARLTS1
UniGene
Hs.558599
Gene Chromosome Location
Chr.13: 49628299-49633872 on Build GRCh38
Transcripts
NM_138450.5 XM_005266253.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4575964 c.299C>T p.A100V Chr.13: 49630746-49630746 on Build GRCh38
2152286 c.369G>A p.K123K Chr.13: 49630816-49630816 on Build GRCh38
6604782 c.455G>A p.R152Q Chr.13: 49630902-49630902 on Build GRCh38
6476093 c.432C>A p.F144L Chr.13: 49630879-49630879 on Build GRCh38
947899 c.454C>T p.R152W Chr.13: 49630901-49630901 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769095684 Chr.13: 49630739-49630739 on Build GRCh38
rs755100942 Chr.13: 49630894-49630894 on Build GRCh38
rs769184018 Chr.13: 49630823-49630823 on Build GRCh38
rs772293845 Chr.13: 49630746-49630746 on Build GRCh38
rs759152923 Chr.13: 49630854-49630854 on Build GRCh38
rs1031176774 Chr.13: 49630822-49630822 on Build GRCh38
rs747335286 Chr.13: 49630916-49630916 on Build GRCh38
rs762961641 Chr.13: 49630946-49630946 on Build GRCh38
rs780101758 Chr.13: 49630738-49630738 on Build GRCh38
rs923965064 Chr.13: 49630872-49630872 on Build GRCh38
rs567358654 Chr.13: 49630891-49630891 on Build GRCh38
rs915492243 Chr.13: 49630902-49630902 on Build GRCh38
rs778849305 Chr.13: 49630901-49630901 on Build GRCh38
rs569508133 Chr.13: 49630936-49630936 on Build GRCh38
rs770655477 Chr.13: 49630844-49630844 on Build GRCh38
rs866872755 Chr.13: 49630827-49630827 on Build GRCh38
rs756352209 Chr.13: 49630882-49630882 on Build GRCh38
rs764627150 Chr.13: 49630858-49630858 on Build GRCh38
rs780120181 Chr.13: 49630802-49630802 on Build GRCh38
rs911644636 Chr.13: 49630941-49630941 on Build GRCh38
rs753334860 Chr.13: 49630772-49630772 on Build GRCh38
rs777416619 Chr.13: 49630784-49630784 on Build GRCh38
rs752542486 Chr.13: 49630778-49630778 on Build GRCh38
rs761431152 Chr.13: 49630781-49630781 on Build GRCh38
rs748581908 Chr.13: 49630833-49630833 on Build GRCh38
rs147120792 Chr.13: 49630839-49630839 on Build GRCh38
rs758766792 Chr.13: 49630907-49630907 on Build GRCh38
rs138452698 Chr.13: 49630843-49630843 on Build GRCh38
rs867095637 Chr.13: 49630895-49630895 on Build GRCh38
rs3803185 Chr.13: 49630889-49630889 on Build GRCh38
rs751003114 Chr.13: 49630840-49630840 on Build GRCh38
rs752737224 Chr.13: 49630859-49630859 on Build GRCh38
rs779514414 Chr.13: 49630832-49630832 on Build GRCh38
rs771389167 Chr.13: 49630921-49630921 on Build GRCh38
rs551214371 Chr.13: 49630927-49630927 on Build GRCh38
rs763952974 Chr.13: 49630878-49630878 on Build GRCh38
rs3803186 Chr.13: 49630744-49630744 on Build GRCh38
rs751295835 Chr.13: 49630880-49630880 on Build GRCh38
rs532945151 Chr.13: 49630775-49630775 on Build GRCh38
rs1051854026 Chr.13: 49630748-49630752 on Build GRCh38
rs748760701 Chr.13: 49630904-49630904 on Build GRCh38
rs970805802 Chr.13: 49630856-49630856 on Build GRCh38
rs766738083 Chr.13: 49630863-49630863 on Build GRCh38
rs756260176 Chr.13: 49630795-49630795 on Build GRCh38
rs772969476 Chr.13: 49630747-49630747 on Build GRCh38
rs762794413 Chr.13: 49630871-49630871 on Build GRCh38
rs776309613 Chr.13: 49630848-49630848 on Build GRCh38
rs868285879 Chr.13: 49630808-49630808 on Build GRCh38
rs760395038 Chr.13: 49630765-49630765 on Build GRCh38
rs35712316 Chr.13: 49630805-49630805 on Build GRCh38
rs759566544 Chr.13: 49630773-49630773 on Build GRCh38
rs745498908 Chr.13: 49630934-49630934 on Build GRCh38
rs994780789 Chr.13: 49630790-49630790 on Build GRCh38
rs766056897 Chr.13: 49630771-49630771 on Build GRCh38
rs34301344 Chr.13: 49630893-49630893 on Build GRCh38
rs778216279 Chr.13: 49630914-49630914 on Build GRCh38
rs750723672 Chr.13: 49630785-49630785 on Build GRCh38
rs551289381 Chr.13: 49630807-49630807 on Build GRCh38
rs774603360 Chr.13: 49630743-49630743 on Build GRCh38
rs146850453 Chr.13: 49630937-49630937 on Build GRCh38

View Full Product Details