Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 210192693-210192966 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACADL
Entrez Gene ID
33
Gene Name
acyl-CoA dehydrogenase, long chain
Gene Aliases
ACAD4, LCAD
UniGene
Hs.471277
Gene Chromosome Location
Chr.2: 210187990-210225491 on Build GRCh38
Transcripts
NM_001608.3 XM_005246517.4 XM_017003955.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1015835 c.1173A>G p.G391G Chr.2: 210192830-210192830 on Build GRCh38
1614244 c.1128A>G p.Q376Q Chr.2: 210192875-210192875 on Build GRCh38
5509122 c.1170G>A p.W390* Chr.2: 210192833-210192833 on Build GRCh38
242768 c.1155G>C p.Q385H Chr.2: 210192848-210192848 on Build GRCh38
4133422 c.1127A>C p.Q376P Chr.2: 210192876-210192876 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746994611 Chr.2: 210192774-210192776 on Build GRCh38
rs755828677 Chr.2: 210192783-210192783 on Build GRCh38
rs749400625 Chr.2: 210192913-210192917 on Build GRCh38
rs780555335 Chr.2: 210192790-210192790 on Build GRCh38
rs895660387 Chr.2: 210192901-210192901 on Build GRCh38
rs779384459 Chr.2: 210192921-210192921 on Build GRCh38
rs755402769 Chr.2: 210192810-210192810 on Build GRCh38
rs996101982 Chr.2: 210192773-210192776 on Build GRCh38
rs140931117 Chr.2: 210192818-210192818 on Build GRCh38
rs888357864 Chr.2: 210192936-210192936 on Build GRCh38
rs753580004 Chr.2: 210192915-210192915 on Build GRCh38
rs762668779 Chr.2: 210192843-210192843 on Build GRCh38
rs922748243 Chr.2: 210192747-210192747 on Build GRCh38
rs776849577 Chr.2: 210192842-210192842 on Build GRCh38
rs769128905 Chr.2: 210192852-210192853 on Build GRCh38
rs771199633 Chr.2: 210192840-210192840 on Build GRCh38
rs993976985 Chr.2: 210192903-210192903 on Build GRCh38
rs755349431 Chr.2: 210192920-210192920 on Build GRCh38
rs867867861 Chr.2: 210192799-210192799 on Build GRCh38
rs777950634 Chr.2: 210192826-210192826 on Build GRCh38
rs544440334 Chr.2: 210192812-210192812 on Build GRCh38
rs17855263 Chr.2: 210192845-210192845 on Build GRCh38
rs374258675 Chr.2: 210192777-210192777 on Build GRCh38
rs760278508 Chr.2: 210192907-210192907 on Build GRCh38
rs753113096 Chr.2: 210192928-210192928 on Build GRCh38
rs761854222 Chr.2: 210192870-210192870 on Build GRCh38
rs561094679 Chr.2: 210192889-210192889 on Build GRCh38
rs368441451 Chr.2: 210192904-210192904 on Build GRCh38
rs766065140 Chr.2: 210192909-210192909 on Build GRCh38
rs766903314 Chr.2: 210192885-210192885 on Build GRCh38
rs781766225 Chr.2: 210192818-210192818 on Build GRCh38
rs767072465 Chr.2: 210192773-210192773 on Build GRCh38
rs915455110 Chr.2: 210192795-210192795 on Build GRCh38
rs1056856525 Chr.2: 210192895-210192895 on Build GRCh38
rs774103658 Chr.2: 210192859-210192859 on Build GRCh38
rs747193831 Chr.2: 210192834-210192834 on Build GRCh38
rs376688105 Chr.2: 210192767-210192767 on Build GRCh38
rs768469986 Chr.2: 210192850-210192850 on Build GRCh38
rs944024241 Chr.2: 210192721-210192721 on Build GRCh38
rs1030776920 Chr.2: 210192857-210192857 on Build GRCh38
rs1041084993 Chr.2: 210192739-210192739 on Build GRCh38

View Full Product Details