Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 15254652-15254925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASB9
Entrez Gene ID
140462
Gene Name
ankyrin repeat and SOCS box containing 9
Gene Aliases
-
UniGene
Hs.19404
Gene Chromosome Location
Chr.X: 15243987-15270476 on Build GRCh38
Transcripts
NM_001031739.2 NM_001168530.1 NM_001168531.1 NM_024087.2 XM_005274446.1 XM_011545458.1 XM_011545460.1 XM_017029283.1 XM_017029284.1 XM_017029285.1 XM_017029286.1

SNP Detail

SNP ID Chromosome Location
rs1041090835 Chr.X: 15254819-15254819 on Build GRCh38
rs2291125 Chr.X: 15254862-15254862 on Build GRCh38
rs751783068 Chr.X: 15254889-15254889 on Build GRCh38
rs900074911 Chr.X: 15254856-15254856 on Build GRCh38
rs752085702 Chr.X: 15254783-15254783 on Build GRCh38
rs746050514 Chr.X: 15254693-15254693 on Build GRCh38
rs144479992 Chr.X: 15254812-15254812 on Build GRCh38
rs780718013 Chr.X: 15254806-15254806 on Build GRCh38
rs750820180 Chr.X: 15254724-15254724 on Build GRCh38
rs761991684 Chr.X: 15254697-15254697 on Build GRCh38
rs372988015 Chr.X: 15254690-15254690 on Build GRCh38
rs747669260 Chr.X: 15254829-15254829 on Build GRCh38
rs368902914 Chr.X: 15254868-15254868 on Build GRCh38
rs745842414 Chr.X: 15254893-15254893 on Build GRCh38
rs867805676 Chr.X: 15254700-15254700 on Build GRCh38
rs184732831 Chr.X: 15254686-15254686 on Build GRCh38
rs769885325 Chr.X: 15254864-15254865 on Build GRCh38
rs376110214 Chr.X: 15254703-15254703 on Build GRCh38
rs189997882 Chr.X: 15254885-15254885 on Build GRCh38
rs943999527 Chr.X: 15254773-15254773 on Build GRCh38
rs766676737 Chr.X: 15254770-15254770 on Build GRCh38
rs764289205 Chr.X: 15254688-15254688 on Build GRCh38
rs755776138 Chr.X: 15254842-15254842 on Build GRCh38
rs755440610 Chr.X: 15254804-15254804 on Build GRCh38
rs777477257 Chr.X: 15254853-15254853 on Build GRCh38
rs780087584 Chr.X: 15254895-15254895 on Build GRCh38
rs200623594 Chr.X: 15254760-15254760 on Build GRCh38
rs1049020013 Chr.X: 15254888-15254888 on Build GRCh38

View Full Product Details