Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 6508851-6509124 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPT2
Entrez Gene ID
285
Gene Name
angiopoietin 2
Gene Aliases
AGPT2, ANG2
UniGene
Hs.583870
Gene Chromosome Location
Chr.8: 6499651-6563420 on Build GRCh38
Transcripts
NM_001118887.1 NM_001118888.1 NM_001147.2 XM_017013318.1 NM_001322042.1 NM_024596.4 XM_011534755.2 XM_011534756.2 XM_011534757.2 XM_011534758.2 XM_011534759.2 XM_011534760.2 XM_017013829.1 XM_017013830.1 XM_017013831.1 XM_017013832.1 XM_017013833.1 XM_017013834.1

Gene Symbol
MCPH1
Entrez Gene ID
79648
Gene Name
microcephalin 1
Gene Aliases
BRIT1, MCT
UniGene
Hs.593807
Gene Chromosome Location
Chr.8: 6406592-6648505 on Build GRCh38
Transcripts
NM_001118887.1 NM_001118888.1 NM_001147.2 XM_017013318.1 NM_001322042.1 NM_024596.4 XM_011534755.2 XM_011534756.2 XM_011534757.2 XM_011534758.2 XM_011534759.2 XM_011534760.2 XM_017013829.1 XM_017013830.1 XM_017013831.1 XM_017013832.1 XM_017013833.1 XM_017013834.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7216123 c.1209T>C p.L403L Chr.8: 6509053-6509053 on Build GRCh38
6472654 c.1232G>A p.G411D Chr.8: 6509030-6509030 on Build GRCh38
1100972 c.1230C>A p.A410A Chr.8: 6509032-6509032 on Build GRCh38
486586 c.1311T>C p.C437C Chr.8: 6508951-6508951 on Build GRCh38
5680245 c.1294A>T p.K432* Chr.8: 6508968-6508968 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148198799 Chr.8: 6508972-6508972 on Build GRCh38
rs773590255 Chr.8: 6509012-6509012 on Build GRCh38
rs759292001 Chr.8: 6509078-6509079 on Build GRCh38
rs749652456 Chr.8: 6509078-6509078 on Build GRCh38
rs754911368 Chr.8: 6508914-6508914 on Build GRCh38
rs753747916 Chr.8: 6508911-6508911 on Build GRCh38
rs777411834 Chr.8: 6508938-6508938 on Build GRCh38
rs750605945 Chr.8: 6508887-6508887 on Build GRCh38
rs367825488 Chr.8: 6509070-6509070 on Build GRCh38
rs138469064 Chr.8: 6509047-6509047 on Build GRCh38
rs769783570 Chr.8: 6508896-6508896 on Build GRCh38
rs751246901 Chr.8: 6509065-6509065 on Build GRCh38
rs767646029 Chr.8: 6508994-6508994 on Build GRCh38
rs760938829 Chr.8: 6509025-6509025 on Build GRCh38
rs977565817 Chr.8: 6508981-6508981 on Build GRCh38
rs748888754 Chr.8: 6508979-6508979 on Build GRCh38
rs200248525 Chr.8: 6508905-6508905 on Build GRCh38
rs963925218 Chr.8: 6508944-6508944 on Build GRCh38
rs948286130 Chr.8: 6509082-6509082 on Build GRCh38
rs774353163 Chr.8: 6508985-6508985 on Build GRCh38
rs746608968 Chr.8: 6508965-6508965 on Build GRCh38
rs554876565 Chr.8: 6509048-6509048 on Build GRCh38
rs770080155 Chr.8: 6508966-6508966 on Build GRCh38
rs769552675 Chr.8: 6509079-6509079 on Build GRCh38
rs370898328 Chr.8: 6508916-6508916 on Build GRCh38
rs896755986 Chr.8: 6509074-6509074 on Build GRCh38
rs61733315 Chr.8: 6509032-6509032 on Build GRCh38
rs946364118 Chr.8: 6509079-6509080 on Build GRCh38
rs777710893 Chr.8: 6508909-6508909 on Build GRCh38
rs57118748 Chr.8: 6509053-6509053 on Build GRCh38
rs1045262552 Chr.8: 6509093-6509093 on Build GRCh38
rs986625651 Chr.8: 6508897-6508897 on Build GRCh38
rs780334639 Chr.8: 6509071-6509071 on Build GRCh38
rs774458437 Chr.8: 6508880-6508880 on Build GRCh38
rs768835261 Chr.8: 6509079-6509079 on Build GRCh38
rs1019549728 Chr.8: 6508928-6508928 on Build GRCh38
rs770728126 Chr.8: 6508876-6508877 on Build GRCh38
rs773106102 Chr.8: 6509066-6509067 on Build GRCh38
rs144041444 Chr.8: 6509055-6509055 on Build GRCh38
rs772309086 Chr.8: 6509100-6509100 on Build GRCh38
rs975259468 Chr.8: 6508945-6508945 on Build GRCh38
rs375795712 Chr.8: 6509095-6509095 on Build GRCh38
rs776523037 Chr.8: 6508993-6508993 on Build GRCh38
rs779574939 Chr.8: 6509090-6509090 on Build GRCh38
rs201230329 Chr.8: 6509037-6509037 on Build GRCh38
rs141563253 Chr.8: 6508993-6508993 on Build GRCh38
rs778332069 Chr.8: 6508915-6508915 on Build GRCh38
rs373034330 Chr.8: 6508881-6508881 on Build GRCh38
rs759447188 Chr.8: 6509035-6509035 on Build GRCh38

View Full Product Details