Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 66020164-66020391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CATSPER1
Entrez Gene ID
117144
Gene Name
cation channel sperm associated 1
Gene Aliases
CATSPER, SPGF7
UniGene
Hs.189105
Gene Chromosome Location
Chr.11: 66016752-66026518 on Build GRCh38
Transcripts
NM_053054.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3752651 c.2070C>T p.A690A Chr.11: 66020195-66020195 on Build GRCh38
378973 c.2071G>A p.A691T Chr.11: 66020194-66020194 on Build GRCh38
5925553 c.2068G>A p.A690T Chr.11: 66020197-66020197 on Build GRCh38
6200341 c.2064+5C>T p.? Chr.11: 66020312-66020312 on Build GRCh38
6894746 c.2047G>C p.E683Q Chr.11: 66020334-66020334 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199668198 Chr.11: 66020282-66020282 on Build GRCh38
rs1004865327 Chr.11: 66020215-66020215 on Build GRCh38
rs745711177 Chr.11: 66020216-66020216 on Build GRCh38
rs369647718 Chr.11: 66020190-66020190 on Build GRCh38
rs773724360 Chr.11: 66020351-66020351 on Build GRCh38
rs34769321 Chr.11: 66020195-66020195 on Build GRCh38
rs201995050 Chr.11: 66020202-66020202 on Build GRCh38
rs530527428 Chr.11: 66020312-66020312 on Build GRCh38
rs1020552356 Chr.11: 66020326-66020326 on Build GRCh38
rs1032676578 Chr.11: 66020284-66020284 on Build GRCh38
rs372660106 Chr.11: 66020307-66020307 on Build GRCh38
rs866087811 Chr.11: 66020322-66020322 on Build GRCh38
rs766690264 Chr.11: 66020185-66020185 on Build GRCh38
rs369285455 Chr.11: 66020194-66020194 on Build GRCh38
rs988402907 Chr.11: 66020343-66020343 on Build GRCh38
rs779811159 Chr.11: 66020227-66020227 on Build GRCh38
rs995833510 Chr.11: 66020228-66020228 on Build GRCh38
rs772556045 Chr.11: 66020350-66020350 on Build GRCh38
rs868337312 Chr.11: 66020370-66020370 on Build GRCh38
rs776914422 Chr.11: 66020287-66020287 on Build GRCh38
rs981253021 Chr.11: 66020371-66020371 on Build GRCh38
rs146534093 Chr.11: 66020316-66020316 on Build GRCh38
rs768246322 Chr.11: 66020237-66020237 on Build GRCh38
rs3829937 Chr.11: 66020195-66020195 on Build GRCh38
rs748682174 Chr.11: 66020234-66020234 on Build GRCh38
rs201528154 Chr.11: 66020217-66020217 on Build GRCh38
rs201385721 Chr.11: 66020274-66020274 on Build GRCh38
rs759492203 Chr.11: 66020295-66020295 on Build GRCh38
rs747724236 Chr.11: 66020332-66020332 on Build GRCh38
rs1028137300 Chr.11: 66020291-66020291 on Build GRCh38
rs143930161 Chr.11: 66020311-66020311 on Build GRCh38
rs762289393 Chr.11: 66020245-66020245 on Build GRCh38
rs752872990 Chr.11: 66020297-66020297 on Build GRCh38
rs748995973 Chr.11: 66020321-66020321 on Build GRCh38
rs755098505 Chr.11: 66020191-66020191 on Build GRCh38
rs12720434 Chr.11: 66020367-66020367 on Build GRCh38
rs141309763 Chr.11: 66020353-66020353 on Build GRCh38
rs3814748 Chr.11: 66020296-66020296 on Build GRCh38
rs774794724 Chr.11: 66020244-66020244 on Build GRCh38
rs766455938 Chr.11: 66020281-66020281 on Build GRCh38
rs955708654 Chr.11: 66020339-66020339 on Build GRCh38
rs750657024 Chr.11: 66020348-66020348 on Build GRCh38
rs373114582 Chr.11: 66020267-66020267 on Build GRCh38
rs754677282 Chr.11: 66020327-66020327 on Build GRCh38
rs772608882 Chr.11: 66020248-66020248 on Build GRCh38
rs376917892 Chr.11: 66020354-66020354 on Build GRCh38
rs778364409 Chr.11: 66020328-66020328 on Build GRCh38

View Full Product Details