Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 74718653-74718912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRDL2
Entrez Gene ID
25884
Gene Name
chordin like 2
Gene Aliases
BNF1, CHL2
UniGene
Hs.432379
Gene Chromosome Location
Chr.11: 74696428-74731385 on Build GRCh38
Transcripts
NM_001278473.2 NM_001304390.1 NM_001304391.1 NM_001304415.1 NM_001304416.1 NM_015424.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5512829 c.127G>A p.G43S Chr.11: 74718788-74718788 on Build GRCh38
4170355 c.118T>G p.Y40D Chr.11: 74718797-74718797 on Build GRCh38
5590203 c.122C>A p.S41Y Chr.11: 74718793-74718793 on Build GRCh38
3398131 c.179G>A p.R60H Chr.11: 74718736-74718736 on Build GRCh38
3453299 c.143C>T p.P48L Chr.11: 74718772-74718772 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765172373 Chr.11: 74718753-74718753 on Build GRCh38
rs942503351 Chr.11: 74718824-74718824 on Build GRCh38
rs200299090 Chr.11: 74718780-74718780 on Build GRCh38
rs763936420 Chr.11: 74718882-74718882 on Build GRCh38
rs779589345 Chr.11: 74718812-74718812 on Build GRCh38
rs775998109 Chr.11: 74718741-74718741 on Build GRCh38
rs748631153 Chr.11: 74718822-74718822 on Build GRCh38
rs756278232 Chr.11: 74718785-74718785 on Build GRCh38
rs568744847 Chr.11: 74718860-74718860 on Build GRCh38
rs1006955208 Chr.11: 74718759-74718759 on Build GRCh38
rs765969607 Chr.11: 74718736-74718736 on Build GRCh38
rs752582264 Chr.11: 74718765-74718765 on Build GRCh38
rs764686642 Chr.11: 74718852-74718852 on Build GRCh38
rs992501750 Chr.11: 74718880-74718880 on Build GRCh38
rs368675432 Chr.11: 74718879-74718879 on Build GRCh38
rs750707329 Chr.11: 74718782-74718782 on Build GRCh38
rs142054086 Chr.11: 74718786-74718786 on Build GRCh38
rs772520244 Chr.11: 74718718-74718718 on Build GRCh38
rs749866447 Chr.11: 74718678-74718678 on Build GRCh38
rs927076168 Chr.11: 74718847-74718847 on Build GRCh38
rs909812691 Chr.11: 74718795-74718795 on Build GRCh38
rs773433183 Chr.11: 74718827-74718827 on Build GRCh38
rs922455545 Chr.11: 74718836-74718836 on Build GRCh38
rs140676656 Chr.11: 74718826-74718826 on Build GRCh38
rs988583183 Chr.11: 74718788-74718788 on Build GRCh38
rs951424322 Chr.11: 74718783-74718783 on Build GRCh38
rs755055015 Chr.11: 74718799-74718799 on Build GRCh38
rs746741046 Chr.11: 74718842-74718842 on Build GRCh38
rs776287863 Chr.11: 74718843-74718843 on Build GRCh38
rs758221620 Chr.11: 74718767-74718767 on Build GRCh38
rs151148265 Chr.11: 74718789-74718789 on Build GRCh38
rs748539674 Chr.11: 74718716-74718716 on Build GRCh38
rs759018807 Chr.11: 74718745-74718745 on Build GRCh38
rs759417155 Chr.11: 74718808-74718808 on Build GRCh38
rs373564234 Chr.11: 74718861-74718861 on Build GRCh38
rs773035564 Chr.11: 74718722-74718722 on Build GRCh38
rs751309267 Chr.11: 74718883-74718883 on Build GRCh38
rs770798372 Chr.11: 74718710-74718717 on Build GRCh38
rs774042578 Chr.11: 74718874-74718874 on Build GRCh38
rs1025592136 Chr.11: 74718761-74718761 on Build GRCh38
rs760219786 Chr.11: 74718735-74718735 on Build GRCh38
rs748995474 Chr.11: 74718693-74718694 on Build GRCh38
rs200715022 Chr.11: 74718773-74718773 on Build GRCh38
rs557854395 Chr.11: 74718694-74718694 on Build GRCh38
rs375388748 Chr.11: 74718702-74718702 on Build GRCh38
rs759069350 Chr.11: 74718850-74718850 on Build GRCh38
rs933745730 Chr.11: 74718874-74718874 on Build GRCh38

View Full Product Details