Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 115956509-115956780 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRK
Entrez Gene ID
2444
Gene Name
fyn related Src family tyrosine kinase
Gene Aliases
GTK, PTK5, RAK
UniGene
Hs.89426
Gene Chromosome Location
Chr.6: 115941031-116100739 on Build GRCh38
Transcripts
NM_002031.2 XM_005266880.4 XM_005266881.2 XM_005266882.4 XM_011535653.2 XM_011535654.2 XM_011535655.2 XM_011535656.2 XM_017010645.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6012440 c.811C>T p.P271S Chr.6: 115956599-115956599 on Build GRCh38
3920609 c.803C>T p.S268L Chr.6: 115956607-115956607 on Build GRCh38
1072343 c.842T>C p.M281T Chr.6: 115956568-115956568 on Build GRCh38
5524384 c.849C>T p.N283N Chr.6: 115956561-115956561 on Build GRCh38
3928041 c.805A>G p.M269V Chr.6: 115956605-115956605 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753688717 Chr.6: 115956570-115956570 on Build GRCh38
rs146859190 Chr.6: 115956595-115956595 on Build GRCh38
rs200172011 Chr.6: 115956605-115956605 on Build GRCh38
rs1048507606 Chr.6: 115956553-115956553 on Build GRCh38
rs570114270 Chr.6: 115956628-115956628 on Build GRCh38
rs776159133 Chr.6: 115956659-115956659 on Build GRCh38
rs746230585 Chr.6: 115956625-115956625 on Build GRCh38
rs765111675 Chr.6: 115956589-115956589 on Build GRCh38
rs771649351 Chr.6: 115956546-115956546 on Build GRCh38
rs553318074 Chr.6: 115956758-115956758 on Build GRCh38
rs748333424 Chr.6: 115956572-115956572 on Build GRCh38
rs759833741 Chr.6: 115956543-115956543 on Build GRCh38
rs17077423 Chr.6: 115956629-115956629 on Build GRCh38
rs755081824 Chr.6: 115956562-115956564 on Build GRCh38
rs1009915678 Chr.6: 115956635-115956635 on Build GRCh38
rs746704835 Chr.6: 115956641-115956641 on Build GRCh38
rs200718000 Chr.6: 115956561-115956561 on Build GRCh38
rs761245233 Chr.6: 115956660-115956660 on Build GRCh38
rs756227805 Chr.6: 115956601-115956601 on Build GRCh38
rs760169315 Chr.6: 115956552-115956552 on Build GRCh38
rs774725821 Chr.6: 115956542-115956542 on Build GRCh38
rs753016460 Chr.6: 115956599-115956599 on Build GRCh38
rs541610371 Chr.6: 115956754-115956754 on Build GRCh38
rs781550644 Chr.6: 115956597-115956597 on Build GRCh38
rs749347146 Chr.6: 115956604-115956604 on Build GRCh38
rs978519675 Chr.6: 115956693-115956693 on Build GRCh38
rs555531595 Chr.6: 115956627-115956627 on Build GRCh38
rs777865782 Chr.6: 115956602-115956602 on Build GRCh38
rs376744061 Chr.6: 115956674-115956674 on Build GRCh38
rs921677197 Chr.6: 115956733-115956733 on Build GRCh38
rs140670243 Chr.6: 115956761-115956761 on Build GRCh38
rs1034677688 Chr.6: 115956647-115956647 on Build GRCh38
rs750164707 Chr.6: 115956592-115956592 on Build GRCh38
rs974017669 Chr.6: 115956678-115956678 on Build GRCh38
rs765214377 Chr.6: 115956661-115956661 on Build GRCh38
rs775194998 Chr.6: 115956550-115956550 on Build GRCh38
rs369393469 Chr.6: 115956554-115956554 on Build GRCh38
rs201455615 Chr.6: 115956646-115956646 on Build GRCh38
rs1023484886 Chr.6: 115956669-115956669 on Build GRCh38

View Full Product Details