Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 41133479-41133752 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBWD6
Entrez Gene ID
644019
Gene Name
COBW domain containing 6
Gene Aliases
-
UniGene
Hs.597638
Gene Chromosome Location
Chr.9: 41131309-41198865 on Build GRCh38
Transcripts
NM_001085457.1 XM_017015019.1 XM_017015020.1 XM_017015021.1 XM_017015022.1 XM_017015023.1 XM_017015024.1 XM_017015025.1 XM_017015026.1 XM_017015028.1 XM_017015029.1 XM_017015031.1 XM_017015033.1 XM_017015034.1 XM_017015035.1 XM_017015036.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4416935 c.521+7T>A p.? Chr.9: 41133498-41133498 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs777244268 Chr.9: 41133672-41133672 on Build GRCh38
rs368998360 Chr.9: 41133687-41133687 on Build GRCh38
rs748985343 Chr.9: 41133683-41133683 on Build GRCh38
rs763969566 Chr.9: 41133705-41133705 on Build GRCh38
rs750860070 Chr.9: 41133651-41133651 on Build GRCh38
rs753945418 Chr.9: 41133706-41133706 on Build GRCh38
rs1008372569 Chr.9: 41133628-41133628 on Build GRCh38
rs770282675 Chr.9: 41133684-41133684 on Build GRCh38
rs747415520 Chr.9: 41133666-41133666 on Build GRCh38
rs780382832 Chr.9: 41133664-41133664 on Build GRCh38
rs769096396 Chr.9: 41133669-41133669 on Build GRCh38
rs772188620 Chr.9: 41133692-41133692 on Build GRCh38
rs774074112 Chr.9: 41133685-41133685 on Build GRCh38
rs762098662 Chr.9: 41133711-41133711 on Build GRCh38
rs373196176 Chr.9: 41133718-41133718 on Build GRCh38
rs62553375 Chr.9: 41133695-41133695 on Build GRCh38
rs542799058 Chr.9: 41133639-41133639 on Build GRCh38
rs199518313 Chr.9: 41133638-41133638 on Build GRCh38
rs757232124 Chr.9: 41133565-41133565 on Build GRCh38
rs760342672 Chr.9: 41133701-41133701 on Build GRCh38
rs758971248 Chr.9: 41133661-41133661 on Build GRCh38

View Full Product Details