Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 49131731-49132002 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM170B
Entrez Gene ID
170370
Gene Name
family with sequence similarity 170 member B
Gene Aliases
C10orf73
UniGene
Hs.646291
Gene Chromosome Location
Chr.10: 49131154-49134020 on Build GRCh38
Transcripts
NM_001164484.1 NR_038973.1

Gene Symbol
FAM170B-AS1
Entrez Gene ID
100506733
Gene Name
FAM170B antisense RNA 1
Gene Aliases
-
UniGene
Hs.404175
Gene Chromosome Location
Chr.10: 49121839-49151547 on Build GRCh38
Transcripts
NM_001164484.1 NR_038973.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5691751 c.667G>C p.A223P Chr.10: 49131798-49131798 on Build GRCh38
3769125 c.579G>A p.P193P Chr.10: 49131886-49131886 on Build GRCh38
918589 c.463G>A p.E155K Chr.10: 49132002-49132002 on Build GRCh38
5551005 c.630C>T p.C210C Chr.10: 49131835-49131835 on Build GRCh38
918588 c.606C>T p.Y202Y Chr.10: 49131859-49131859 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs561798621 Chr.10: 49131900-49131900 on Build GRCh38
rs747326148 Chr.10: 49131891-49131891 on Build GRCh38
rs940024733 Chr.10: 49131883-49131883 on Build GRCh38
rs759154687 Chr.10: 49131766-49131766 on Build GRCh38
rs754239909 Chr.10: 49131852-49131852 on Build GRCh38
rs759103665 Chr.10: 49131940-49131940 on Build GRCh38
rs1018573310 Chr.10: 49131854-49131854 on Build GRCh38
rs75297145 Chr.10: 49131917-49131917 on Build GRCh38
rs920627125 Chr.10: 49131813-49131813 on Build GRCh38
rs968309293 Chr.10: 49131765-49131765 on Build GRCh38
rs529073950 Chr.10: 49131904-49131904 on Build GRCh38
rs900252567 Chr.10: 49131898-49131898 on Build GRCh38
rs767251917 Chr.10: 49131784-49131784 on Build GRCh38
rs373146727 Chr.10: 49131851-49131851 on Build GRCh38
rs749138610 Chr.10: 49131909-49131909 on Build GRCh38
rs756429385 Chr.10: 49131818-49131818 on Build GRCh38
rs757786364 Chr.10: 49131853-49131853 on Build GRCh38
rs746362171 Chr.10: 49131763-49131769 on Build GRCh38
rs907016362 Chr.10: 49131965-49131965 on Build GRCh38
rs745916600 Chr.10: 49131872-49131872 on Build GRCh38
rs979561254 Chr.10: 49131786-49131786 on Build GRCh38
rs778926356 Chr.10: 49131869-49131869 on Build GRCh38
rs781753059 Chr.10: 49131832-49131832 on Build GRCh38
rs908020603 Chr.10: 49131868-49131868 on Build GRCh38
rs780131272 Chr.10: 49131887-49131887 on Build GRCh38
rs866574926 Chr.10: 49131858-49131858 on Build GRCh38
rs1021163545 Chr.10: 49131758-49131758 on Build GRCh38
rs73302791 Chr.10: 49131925-49131925 on Build GRCh38
rs12765281 Chr.10: 49131880-49131880 on Build GRCh38
rs1040115705 Chr.10: 49131893-49131893 on Build GRCh38
rs972638069 Chr.10: 49131901-49131901 on Build GRCh38
rs752547553 Chr.10: 49131968-49131968 on Build GRCh38
rs752452944 Chr.10: 49131814-49131814 on Build GRCh38
rs1020175450 Chr.10: 49131949-49131949 on Build GRCh38
rs992670815 Chr.10: 49131864-49131864 on Build GRCh38

View Full Product Details