Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 152309752-152310011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLG
Entrez Gene ID
2312
Gene Name
filaggrin
Gene Aliases
ATOD2
UniGene
Hs.654510
Gene Chromosome Location
Chr.1: 152302175-152325203 on Build GRCh38
Transcripts
NM_002016.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5661327 c.5090C>A p.T1697N Chr.1: 152309796-152309796 on Build GRCh38
7257486 c.5080G>C p.D1694H Chr.1: 152309806-152309806 on Build GRCh38
1667969 c.5096G>A p.R1699H Chr.1: 152309790-152309790 on Build GRCh38
3474030 c.5033G>A p.G1678E Chr.1: 152309853-152309853 on Build GRCh38
5733179 c.5040A>C p.R1680S Chr.1: 152309846-152309846 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752102769 Chr.1: 152309784-152309784 on Build GRCh38
rs749542190 Chr.1: 152309862-152309862 on Build GRCh38
rs142493095 Chr.1: 152309962-152309962 on Build GRCh38
rs12407807 Chr.1: 152309835-152309835 on Build GRCh38
rs760059994 Chr.1: 152309991-152309991 on Build GRCh38
rs555631824 Chr.1: 152309890-152309890 on Build GRCh38
rs560229190 Chr.1: 152309959-152309959 on Build GRCh38
rs201986758 Chr.1: 152309979-152309979 on Build GRCh38
rs773467871 Chr.1: 152309776-152309776 on Build GRCh38
rs12405278 Chr.1: 152309791-152309791 on Build GRCh38
rs776793261 Chr.1: 152309923-152309923 on Build GRCh38
rs760100930 Chr.1: 152309816-152309816 on Build GRCh38
rs542448683 Chr.1: 152309888-152309888 on Build GRCh38
rs373806639 Chr.1: 152309857-152309857 on Build GRCh38
rs764106518 Chr.1: 152309976-152309976 on Build GRCh38
rs776060660 Chr.1: 152309949-152309949 on Build GRCh38
rs758146665 Chr.1: 152309988-152309988 on Build GRCh38
rs936961132 Chr.1: 152309990-152309990 on Build GRCh38
rs151252691 Chr.1: 152309936-152309936 on Build GRCh38
rs764236927 Chr.1: 152309830-152309830 on Build GRCh38
rs763599249 Chr.1: 152309954-152309954 on Build GRCh38
rs775926509 Chr.1: 152309920-152309920 on Build GRCh38
rs774573308 Chr.1: 152309878-152309878 on Build GRCh38
rs144090343 Chr.1: 152309800-152309800 on Build GRCh38
rs751543259 Chr.1: 152309837-152309837 on Build GRCh38
rs760723558 Chr.1: 152309873-152309873 on Build GRCh38
rs542735959 Chr.1: 152309941-152309941 on Build GRCh38
rs780024568 Chr.1: 152309793-152309793 on Build GRCh38
rs746942219 Chr.1: 152309794-152309794 on Build GRCh38
rs772851618 Chr.1: 152309802-152309802 on Build GRCh38
rs752678129 Chr.1: 152309907-152309907 on Build GRCh38
rs777358702 Chr.1: 152309853-152309853 on Build GRCh38
rs200129570 Chr.1: 152309957-152309957 on Build GRCh38
rs558022638 Chr.1: 152309777-152309777 on Build GRCh38
rs766130240 Chr.1: 152309836-152309836 on Build GRCh38
rs769874552 Chr.1: 152309810-152309810 on Build GRCh38
rs757002494 Chr.1: 152309915-152309915 on Build GRCh38
rs756314779 Chr.1: 152309788-152309788 on Build GRCh38
rs373818541 Chr.1: 152309967-152309967 on Build GRCh38
rs139975241 Chr.1: 152309919-152309919 on Build GRCh38
rs754076126 Chr.1: 152309834-152309834 on Build GRCh38
rs761483019 Chr.1: 152309984-152309984 on Build GRCh38
rs775745183 Chr.1: 152309824-152309824 on Build GRCh38
rs753393394 Chr.1: 152309948-152309948 on Build GRCh38
rs753707307 Chr.1: 152309977-152309977 on Build GRCh38
rs771327271 Chr.1: 152309808-152309808 on Build GRCh38
rs779532903 Chr.1: 152309989-152309989 on Build GRCh38
rs201990594 Chr.1: 152309911-152309911 on Build GRCh38
rs758153809 Chr.1: 152309792-152309792 on Build GRCh38
rs201910898 Chr.1: 152309917-152309917 on Build GRCh38
rs905562362 Chr.1: 152309782-152309782 on Build GRCh38
rs769848573 Chr.1: 152309964-152309964 on Build GRCh38
rs764228164 Chr.1: 152309876-152309876 on Build GRCh38
rs778766819 Chr.1: 152309953-152309953 on Build GRCh38
rs772712398 Chr.1: 152309865-152309865 on Build GRCh38
rs370872995 Chr.1: 152309963-152309963 on Build GRCh38
rs140491110 Chr.1: 152309950-152309950 on Build GRCh38
rs752654274 Chr.1: 152309969-152309969 on Build GRCh38
rs775390438 Chr.1: 152309970-152309970 on Build GRCh38
rs778016068 Chr.1: 152309790-152309790 on Build GRCh38
rs770005572 Chr.1: 152309935-152309935 on Build GRCh38
rs767453412 Chr.1: 152309906-152309906 on Build GRCh38
rs776084236 Chr.1: 152309869-152309869 on Build GRCh38
rs139318679 Chr.1: 152309985-152309985 on Build GRCh38
rs768090281 Chr.1: 152309823-152309823 on Build GRCh38
rs763289326 Chr.1: 152309804-152309804 on Build GRCh38
rs376694731 Chr.1: 152309849-152309849 on Build GRCh38
rs760743145 Chr.1: 152309827-152309827 on Build GRCh38
rs754699349 Chr.1: 152309896-152309896 on Build GRCh38
rs200033409 Chr.1: 152309908-152309908 on Build GRCh38
rs201238283 Chr.1: 152309841-152309841 on Build GRCh38
rs746195146 Chr.1: 152309864-152309864 on Build GRCh38
rs771090956 Chr.1: 152309872-152309872 on Build GRCh38
rs761754522 Chr.1: 152309924-152309924 on Build GRCh38
rs768667496 Chr.1: 152309922-152309922 on Build GRCh38
rs377260223 Chr.1: 152309958-152309958 on Build GRCh38
rs1016662551 Chr.1: 152309856-152309856 on Build GRCh38
rs778105843 Chr.1: 152309835-152309835 on Build GRCh38
rs780654991 Chr.1: 152309798-152309798 on Build GRCh38
rs760321940 Chr.1: 152309975-152309975 on Build GRCh38
rs748700632 Chr.1: 152309909-152309909 on Build GRCh38
rs748041147 Chr.1: 152309807-152309808 on Build GRCh38
rs148468221 Chr.1: 152309843-152309843 on Build GRCh38
rs768193909 Chr.1: 152309796-152309796 on Build GRCh38
rs533186028 Chr.1: 152309942-152309942 on Build GRCh38
rs200684284 Chr.1: 152309799-152309799 on Build GRCh38
rs751383400 Chr.1: 152309893-152309893 on Build GRCh38
rs145738429 Chr.1: 152309785-152309785 on Build GRCh38
rs760688738 Chr.1: 152309947-152309947 on Build GRCh38
rs766812472 Chr.1: 152309781-152309781 on Build GRCh38
rs754927316 Chr.1: 152309838-152309838 on Build GRCh38

View Full Product Details