Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 74344889-74345154 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP11A1
Entrez Gene ID
1583
Gene Name
cytochrome P450 family 11 subfamily A member 1
Gene Aliases
CYP11A, CYPXIA1, P450SCC
UniGene
Hs.303980
Gene Chromosome Location
Chr.15: 74337762-74367740 on Build GRCh38
Transcripts
NM_000781.2 NM_001099773.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5342623 c.526C>T p.R176W Chr.15: 74345143-74345143 on Build GRCh38
1301414 c.553C>T p.R185C Chr.15: 74345116-74345116 on Build GRCh38
1708394 c.572C>T p.S191F Chr.15: 74345097-74345097 on Build GRCh38
5599368 c.529G>A p.D177N Chr.15: 74345140-74345140 on Build GRCh38
3503921 c.586G>A p.G196R Chr.15: 74345083-74345083 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762299364 Chr.15: 74345124-74345124 on Build GRCh38
rs764579087 Chr.15: 74345031-74345031 on Build GRCh38
rs777958141 Chr.15: 74344983-74344993 on Build GRCh38
rs759322669 Chr.15: 74345100-74345100 on Build GRCh38
rs759720679 Chr.15: 74344983-74344983 on Build GRCh38
rs775659306 Chr.15: 74345114-74345114 on Build GRCh38
rs200235389 Chr.15: 74345129-74345129 on Build GRCh38
rs771857709 Chr.15: 74344965-74344965 on Build GRCh38
rs121912811 Chr.15: 74345103-74345103 on Build GRCh38
rs774045073 Chr.15: 74344943-74344943 on Build GRCh38
rs746888440 Chr.15: 74345087-74345087 on Build GRCh38
rs576925843 Chr.15: 74345085-74345085 on Build GRCh38
rs371041903 Chr.15: 74345083-74345083 on Build GRCh38
rs202119174 Chr.15: 74345056-74345056 on Build GRCh38
rs747738719 Chr.15: 74344952-74344952 on Build GRCh38
rs1053474473 Chr.15: 74344973-74344973 on Build GRCh38
rs886051480 Chr.15: 74345080-74345080 on Build GRCh38
rs150405116 Chr.15: 74345135-74345135 on Build GRCh38
rs1008502067 Chr.15: 74345081-74345081 on Build GRCh38
rs764446555 Chr.15: 74345116-74345116 on Build GRCh38
rs74522504 Chr.15: 74344921-74344921 on Build GRCh38
rs760551103 Chr.15: 74344977-74344977 on Build GRCh38
rs58174009 Chr.15: 74345134-74345134 on Build GRCh38
rs763099365 Chr.15: 74345115-74345115 on Build GRCh38
rs1028546507 Chr.15: 74345051-74345051 on Build GRCh38
rs752975205 Chr.15: 74345008-74345008 on Build GRCh38
rs781754675 Chr.15: 74345058-74345058 on Build GRCh38
rs1027076275 Chr.15: 74345117-74345117 on Build GRCh38
rs555219883 Chr.15: 74344949-74344949 on Build GRCh38
rs776643277 Chr.15: 74345096-74345096 on Build GRCh38
rs146892548 Chr.15: 74345095-74345095 on Build GRCh38
rs746351603 Chr.15: 74345059-74345059 on Build GRCh38
rs772949596 Chr.15: 74344969-74344969 on Build GRCh38
rs900137598 Chr.15: 74345023-74345023 on Build GRCh38
rs572665499 Chr.15: 74344920-74344920 on Build GRCh38
rs776428826 Chr.15: 74344981-74344981 on Build GRCh38
rs376978027 Chr.15: 74344911-74344911 on Build GRCh38
rs776068433 Chr.15: 74345053-74345053 on Build GRCh38
rs536464242 Chr.15: 74345109-74345109 on Build GRCh38
rs766387844 Chr.15: 74344979-74344979 on Build GRCh38
rs112977174 Chr.15: 74345029-74345029 on Build GRCh38
rs770022852 Chr.15: 74345102-74345102 on Build GRCh38
rs771665524 Chr.15: 74345084-74345084 on Build GRCh38
rs576390546 Chr.15: 74344986-74344986 on Build GRCh38
rs778329815 Chr.15: 74345062-74345062 on Build GRCh38

View Full Product Details