Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 33237833-33238066 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BPIFA1
Entrez Gene ID
51297
Gene Name
BPI fold containing family A member 1
Gene Aliases
LUNX, NASG, PLUNC, SPLUNC1, SPURT, bA49G10.5
UniGene
Hs.211092
Gene Chromosome Location
Chr.20: 33235996-33243309 on Build GRCh38
Transcripts
NM_001243193.1 NM_016583.3 NM_130852.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6612325 c.155C>T p.T52I Chr.20: 33237866-33237866 on Build GRCh38
5777514 c.139C>G p.L47V Chr.20: 33237850-33237850 on Build GRCh38
5365869 c.159T>C p.N53N Chr.20: 33237870-33237870 on Build GRCh38
577107 c.147A>G p.G49G Chr.20: 33237858-33237858 on Build GRCh38
6873287 c.167G>A p.S56N Chr.20: 33238061-33238061 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765059597 Chr.20: 33237890-33237890 on Build GRCh38
rs369322707 Chr.20: 33238005-33238005 on Build GRCh38
rs757636341 Chr.20: 33237899-33237916 on Build GRCh38
rs190416867 Chr.20: 33237967-33237967 on Build GRCh38
rs902016822 Chr.20: 33237950-33237950 on Build GRCh38
rs185078840 Chr.20: 33237860-33237860 on Build GRCh38
rs1044169577 Chr.20: 33237914-33237914 on Build GRCh38
rs539781988 Chr.20: 33237885-33237885 on Build GRCh38
rs984723384 Chr.20: 33237898-33237898 on Build GRCh38
rs2889735 Chr.20: 33237907-33237907 on Build GRCh38
rs907881048 Chr.20: 33237909-33237909 on Build GRCh38
rs745886294 Chr.20: 33237899-33237910 on Build GRCh38
rs778166139 Chr.20: 33237898-33237911 on Build GRCh38
rs771170398 Chr.20: 33237864-33237864 on Build GRCh38
rs1054102281 Chr.20: 33237921-33237921 on Build GRCh38
rs759112850 Chr.20: 33237898-33237909 on Build GRCh38
rs1407019 Chr.20: 33237991-33237991 on Build GRCh38
rs113886978 Chr.20: 33237882-33238142 on Build GRCh38
rs762527387 Chr.20: 33237898-33237907 on Build GRCh38
rs777446931 Chr.20: 33237898-33237913 on Build GRCh38
rs60280161 Chr.20: 33237944-33237951 on Build GRCh38
rs952102562 Chr.20: 33237974-33237974 on Build GRCh38
rs796552647 Chr.20: 33237897-33237898 on Build GRCh38
rs780401811 Chr.20: 33237899-33237912 on Build GRCh38
rs962385031 Chr.20: 33237997-33237997 on Build GRCh38
rs973085650 Chr.20: 33237911-33237911 on Build GRCh38
rs539893446 Chr.20: 33237899-33237908 on Build GRCh38
rs777514813 Chr.20: 33237899-33237899 on Build GRCh38
rs749582671 Chr.20: 33238026-33238026 on Build GRCh38
rs772043447 Chr.20: 33237889-33237898 on Build GRCh38
rs2889736 Chr.20: 33237908-33237908 on Build GRCh38
rs556710653 Chr.20: 33238027-33238027 on Build GRCh38
rs768246418 Chr.20: 33237899-33237900 on Build GRCh38
rs772781238 Chr.20: 33237909-33237924 on Build GRCh38
rs1020667679 Chr.20: 33237996-33237996 on Build GRCh38
rs905575004 Chr.20: 33237915-33237915 on Build GRCh38
rs751255503 Chr.20: 33237905-33237905 on Build GRCh38
rs766593708 Chr.20: 33237905-33237914 on Build GRCh38
rs773271112 Chr.20: 33237890-33237897 on Build GRCh38
rs780438901 Chr.20: 33237899-33237914 on Build GRCh38
rs199865803 Chr.20: 33238010-33238010 on Build GRCh38
rs138768031 Chr.20: 33237978-33237978 on Build GRCh38
rs747257829 Chr.20: 33238024-33238024 on Build GRCh38
rs762642615 Chr.20: 33237901-33237901 on Build GRCh38
rs752101500 Chr.20: 33237899-33237906 on Build GRCh38
rs935746802 Chr.20: 33237916-33237916 on Build GRCh38
rs1047880651 Chr.20: 33237857-33237857 on Build GRCh38
rs57665702 Chr.20: 33237938-33237951 on Build GRCh38
rs906122003 Chr.20: 33237919-33237919 on Build GRCh38
rs755543961 Chr.20: 33237920-33237920 on Build GRCh38
rs60513860 Chr.20: 33237936-33237951 on Build GRCh38
rs774139260 Chr.20: 33237907-33237914 on Build GRCh38
rs939931063 Chr.20: 33237910-33237910 on Build GRCh38
rs761669287 Chr.20: 33237889-33237890 on Build GRCh38
rs760599937 Chr.20: 33237893-33237898 on Build GRCh38
rs535531740 Chr.20: 33238019-33238019 on Build GRCh38
rs776858314 Chr.20: 33238038-33238038 on Build GRCh38
rs949900499 Chr.20: 33237913-33237913 on Build GRCh38
rs759560502 Chr.20: 33237888-33237888 on Build GRCh38
rs917077575 Chr.20: 33237912-33237912 on Build GRCh38
rs748512574 Chr.20: 33238007-33238007 on Build GRCh38
rs763692102 Chr.20: 33237903-33237903 on Build GRCh38
rs1044645094 Chr.20: 33237900-33237900 on Build GRCh38
rs534398489 Chr.20: 33237980-33237980 on Build GRCh38
rs754273978 Chr.20: 33237917-33237917 on Build GRCh38
rs60370165 Chr.20: 33237946-33237951 on Build GRCh38
rs1019646983 Chr.20: 33237944-33237944 on Build GRCh38
rs891477385 Chr.20: 33237922-33237922 on Build GRCh38
rs1010473724 Chr.20: 33237936-33237936 on Build GRCh38
rs764826086 Chr.20: 33237898-33237903 on Build GRCh38
rs11468029 Chr.20: 33237942-33237951 on Build GRCh38
rs1029125072 Chr.20: 33237970-33237970 on Build GRCh38
rs745797507 Chr.20: 33238046-33238046 on Build GRCh38
rs766346818 Chr.20: 33237897-33237898 on Build GRCh38
rs75416632 Chr.20: 33237906-33237906 on Build GRCh38

View Full Product Details