Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44913841-44914082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFAP
Entrez Gene ID
2670
Gene Name
glial fibrillary acidic protein
Gene Aliases
ALXDRD
UniGene
Hs.514227
Gene Chromosome Location
Chr.17: 44905626-44915552 on Build GRCh38
Transcripts
NM_001131019.2 NM_001242376.1 NM_002055.4 XM_017024451.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6254036 c.522+3C>A p.? Chr.17: 44914025-44914025 on Build GRCh38
4627540 c.482T>C p.L161P Chr.17: 44914068-44914068 on Build GRCh38
4627541 c.482T>C p.L161P Chr.17: 44914068-44914068 on Build GRCh38
4631639 c.498G>A p.E166E Chr.17: 44914052-44914052 on Build GRCh38
4631640 c.498G>A p.E166E Chr.17: 44914052-44914052 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768627851 Chr.17: 44913871-44913871 on Build GRCh38
rs778488589 Chr.17: 44914015-44914015 on Build GRCh38
rs908566085 Chr.17: 44914003-44914003 on Build GRCh38
rs760384361 Chr.17: 44914002-44914002 on Build GRCh38
rs975648481 Chr.17: 44913930-44913930 on Build GRCh38
rs1010495954 Chr.17: 44914001-44914001 on Build GRCh38
rs1045884031 Chr.17: 44913924-44913924 on Build GRCh38
rs931406444 Chr.17: 44913945-44913945 on Build GRCh38
rs759448342 Chr.17: 44913979-44913979 on Build GRCh38
rs903378821 Chr.17: 44913942-44913942 on Build GRCh38
rs267607668 Chr.17: 44913899-44913899 on Build GRCh38
rs78616766 Chr.17: 44914060-44914060 on Build GRCh38
rs1031147034 Chr.17: 44913958-44913958 on Build GRCh38
rs569658813 Chr.17: 44913965-44913965 on Build GRCh38
rs761298500 Chr.17: 44913982-44913982 on Build GRCh38
rs775293988 Chr.17: 44913864-44913864 on Build GRCh38
rs570491117 Chr.17: 44914016-44914016 on Build GRCh38
rs781332429 Chr.17: 44914036-44914036 on Build GRCh38
rs893561119 Chr.17: 44913969-44913969 on Build GRCh38
rs368066315 Chr.17: 44914012-44914012 on Build GRCh38
rs770076335 Chr.17: 44914054-44914054 on Build GRCh38
rs1047105744 Chr.17: 44913959-44913959 on Build GRCh38
rs143583428 Chr.17: 44914034-44914034 on Build GRCh38
rs552120023 Chr.17: 44914013-44914013 on Build GRCh38
rs774243225 Chr.17: 44913980-44913980 on Build GRCh38
rs749845205 Chr.17: 44913996-44913996 on Build GRCh38
rs780225821 Chr.17: 44914062-44914062 on Build GRCh38
rs34001162 Chr.17: 44914023-44914023 on Build GRCh38
rs920056947 Chr.17: 44913944-44913944 on Build GRCh38
rs769841772 Chr.17: 44913863-44913863 on Build GRCh38
rs530706383 Chr.17: 44914007-44914007 on Build GRCh38
rs767041749 Chr.17: 44913992-44913992 on Build GRCh38
rs745894599 Chr.17: 44914046-44914046 on Build GRCh38

View Full Product Details