Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 47137195-47137442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC27
Entrez Gene ID
996
Gene Name
cell division cycle 27
Gene Aliases
ANAPC3, APC3, CDC27Hs, D0S1430E, D17S978E, H-NUC, HNUC, NUC2
UniGene
Hs.463295
Gene Chromosome Location
Chr.17: 47117697-47189332 on Build GRCh38
Transcripts
NM_001114091.2 NM_001256.4 NM_001293089.1 NM_001293091.1 XM_011525546.2 XM_011525547.2 XM_011525548.2 XM_011525549.2 XM_011525550.2 XM_017025481.1 XM_017025482.1 XM_017025483.1 XM_017025484.1 XM_017025485.1 XM_017025486.1 XM_017025487.1 XM_017025488.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4130187 c.1857A>C p.L619F Chr.17: 47137226-47137226 on Build GRCh38
980365 c.1864G>T p.E622* Chr.17: 47137219-47137219 on Build GRCh38
1721173 c.1768G>A p.D590N Chr.17: 47137315-47137315 on Build GRCh38
1563799 c.1808C>T p.A603V Chr.17: 47137257-47137257 on Build GRCh38
4268063 c.1843C>A p.H615N Chr.17: 47137240-47137240 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11570544 Chr.17: 47137265-47137265 on Build GRCh38
rs749015475 Chr.17: 47137355-47137355 on Build GRCh38
rs757498494 Chr.17: 47137362-47137363 on Build GRCh38
rs200907637 Chr.17: 47137332-47137332 on Build GRCh38
rs765426042 Chr.17: 47137327-47137327 on Build GRCh38
rs761249462 Chr.17: 47137370-47137370 on Build GRCh38
rs11570543 Chr.17: 47137319-47137319 on Build GRCh38
rs749723942 Chr.17: 47137363-47137369 on Build GRCh38
rs74390782 Chr.17: 47137324-47137324 on Build GRCh38
rs752977065 Chr.17: 47137328-47137328 on Build GRCh38
rs765661411 Chr.17: 47137381-47137381 on Build GRCh38
rs536202280 Chr.17: 47137376-47137376 on Build GRCh38
rs746915890 Chr.17: 47137363-47137364 on Build GRCh38
rs779126942 Chr.17: 47137247-47137247 on Build GRCh38
rs138020641 Chr.17: 47137277-47137277 on Build GRCh38
rs75637741 Chr.17: 47137257-47137257 on Build GRCh38
rs764881133 Chr.17: 47137359-47137360 on Build GRCh38
rs77871715 Chr.17: 47137363-47137363 on Build GRCh38
rs776229474 Chr.17: 47137372-47137372 on Build GRCh38
rs748198995 Chr.17: 47137254-47137254 on Build GRCh38
rs796427812 Chr.17: 47137248-47137248 on Build GRCh38
rs201293466 Chr.17: 47137342-47137342 on Build GRCh38
rs755364658 Chr.17: 47137325-47137325 on Build GRCh38
rs148087250 Chr.17: 47137267-47137267 on Build GRCh38
rs62075623 Chr.17: 47137333-47137333 on Build GRCh38
rs79285536 Chr.17: 47137226-47137226 on Build GRCh38
rs766866034 Chr.17: 47137377-47137377 on Build GRCh38
rs111322439 Chr.17: 47137295-47137295 on Build GRCh38
rs773586774 Chr.17: 47137310-47137310 on Build GRCh38
rs796265930 Chr.17: 47137285-47137285 on Build GRCh38
rs775321736 Chr.17: 47137288-47137288 on Build GRCh38
rs76926116 Chr.17: 47137239-47137239 on Build GRCh38
rs796561781 Chr.17: 47137249-47137249 on Build GRCh38
rs75661039 Chr.17: 47137238-47137238 on Build GRCh38
rs534711882 Chr.17: 47137264-47137264 on Build GRCh38
rs763754940 Chr.17: 47137308-47137308 on Build GRCh38
rs200200993 Chr.17: 47137270-47137270 on Build GRCh38
rs769759034 Chr.17: 47137348-47137348 on Build GRCh38
rs62075621 Chr.17: 47137307-47137307 on Build GRCh38
rs200389111 Chr.17: 47137408-47137408 on Build GRCh38
rs199588106 Chr.17: 47137360-47137360 on Build GRCh38
rs758473592 Chr.17: 47137330-47137330 on Build GRCh38
rs781023793 Chr.17: 47137343-47137343 on Build GRCh38
rs761441999 Chr.17: 47137282-47137282 on Build GRCh38
rs138264973 Chr.17: 47137251-47137252 on Build GRCh38
rs62075620 Chr.17: 47137303-47137303 on Build GRCh38
rs749867179 Chr.17: 47137361-47137362 on Build GRCh38
rs777678685 Chr.17: 47137260-47137260 on Build GRCh38
rs147346652 Chr.17: 47137364-47137364 on Build GRCh38
rs796222654 Chr.17: 47137250-47137250 on Build GRCh38
rs62075622 Chr.17: 47137316-47137316 on Build GRCh38
rs796359371 Chr.17: 47137251-47137251 on Build GRCh38
rs769532884 Chr.17: 47137280-47137280 on Build GRCh38
rs79260965 Chr.17: 47137240-47137240 on Build GRCh38
rs796969472 Chr.17: 47137282-47137282 on Build GRCh38
rs112627286 Chr.17: 47137234-47137234 on Build GRCh38
rs775268664 Chr.17: 47137349-47137349 on Build GRCh38

View Full Product Details