Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 40021836-40022061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM83F
Entrez Gene ID
113828
Gene Name
family with sequence similarity 83 member F
Gene Aliases
-
UniGene
Hs.197680
Gene Chromosome Location
Chr.22: 39994949-40030039 on Build GRCh38
Transcripts
NM_138435.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4683258 c.1355C>T p.A452V Chr.22: 40021865-40021865 on Build GRCh38
3554789 c.1329C>T p.F443F Chr.22: 40021839-40021839 on Build GRCh38
3912871 c.1429G>A p.E477K Chr.22: 40021939-40021939 on Build GRCh38
4104356 c.1440T>C p.S480S Chr.22: 40021950-40021950 on Build GRCh38
4104355 c.1369G>A p.A457T Chr.22: 40021879-40021879 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770039552 Chr.22: 40021935-40021935 on Build GRCh38
rs747127782 Chr.22: 40021917-40021917 on Build GRCh38
rs1018618515 Chr.22: 40022022-40022022 on Build GRCh38
rs755112704 Chr.22: 40021997-40021997 on Build GRCh38
rs894210091 Chr.22: 40021857-40021857 on Build GRCh38
rs41276305 Chr.22: 40021939-40021939 on Build GRCh38
rs778728277 Chr.22: 40021879-40021879 on Build GRCh38
rs751413182 Chr.22: 40021990-40021990 on Build GRCh38
rs370605308 Chr.22: 40021909-40021909 on Build GRCh38
rs760259740 Chr.22: 40021946-40021946 on Build GRCh38
rs189817068 Chr.22: 40021867-40021867 on Build GRCh38
rs752311146 Chr.22: 40021862-40021862 on Build GRCh38
rs201189104 Chr.22: 40021960-40021960 on Build GRCh38
rs552981943 Chr.22: 40021938-40021938 on Build GRCh38
rs145215263 Chr.22: 40021866-40021866 on Build GRCh38
rs763459853 Chr.22: 40021953-40021953 on Build GRCh38
rs755659577 Chr.22: 40022018-40022018 on Build GRCh38
rs553725529 Chr.22: 40021896-40021896 on Build GRCh38
rs199806449 Chr.22: 40021871-40021871 on Build GRCh38
rs761411099 Chr.22: 40021959-40021959 on Build GRCh38
rs147178151 Chr.22: 40021923-40021923 on Build GRCh38
rs755519348 Chr.22: 40021865-40021865 on Build GRCh38
rs1015529957 Chr.22: 40021905-40021905 on Build GRCh38
rs779971762 Chr.22: 40021981-40021981 on Build GRCh38
rs768553111 Chr.22: 40021918-40021918 on Build GRCh38
rs945581746 Chr.22: 40021956-40021956 on Build GRCh38
rs753366673 Chr.22: 40021955-40021955 on Build GRCh38
rs376593839 Chr.22: 40021984-40021984 on Build GRCh38
rs781508185 Chr.22: 40021922-40021922 on Build GRCh38
rs915355558 Chr.22: 40021952-40021952 on Build GRCh38
rs376939521 Chr.22: 40021897-40021897 on Build GRCh38
rs1014828410 Chr.22: 40021948-40021948 on Build GRCh38
rs1012713304 Chr.22: 40021863-40021863 on Build GRCh38
rs750278455 Chr.22: 40021965-40021965 on Build GRCh38
rs541080782 Chr.22: 40021937-40021937 on Build GRCh38
rs758133724 Chr.22: 40021968-40021968 on Build GRCh38
rs1026620018 Chr.22: 40021998-40021998 on Build GRCh38
rs965358820 Chr.22: 40022023-40022023 on Build GRCh38
rs779955287 Chr.22: 40021907-40021907 on Build GRCh38
rs976554365 Chr.22: 40022033-40022033 on Build GRCh38
rs952500292 Chr.22: 40022013-40022013 on Build GRCh38
rs563529855 Chr.22: 40022039-40022039 on Build GRCh38
rs774928948 Chr.22: 40021941-40021941 on Build GRCh38

View Full Product Details