Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161173824-161174081 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALT3
Entrez Gene ID
8703
Gene Name
beta-1,4-galactosyltransferase 3
Gene Aliases
beta4Gal-T3
UniGene
Hs.321231
Gene Chromosome Location
Chr.1: 161171310-161177968 on Build GRCh38
Transcripts
NM_001199873.1 NM_001199874.1 NM_003779.3 XM_005245566.1 XM_011510093.1 XM_017002714.1 XM_017002715.1 XM_005245291.3 XM_005245295.3 XM_006711402.2 XM_006711403.2 XM_006711406.2 XM_011509663.2 XM_011509664.1 XM_011509665.2 XM_011509666.2 XM_011509667.2 XM_011509668.2 XM_011509669.1 XM_011509670.2 XM_011509673.2 XM_011509674.2 XM_011509676.1 XM_011509677.1 XM_011509678.1 XM_011509679.1 XM_011509681.1 XM_011509682.1 XM_017001561.1 XM_017001562.1 XM_017001563.1 XM_017001564.1 XM_017001570.1 XM_017001571.1

Gene Symbol
PPOX
Entrez Gene ID
5498
Gene Name
protoporphyrinogen oxidase
Gene Aliases
PPO, V290M, VP
UniGene
Hs.517373
Gene Chromosome Location
Chr.1: 161165728-161178277 on Build GRCh38
Transcripts
NM_001199873.1 NM_001199874.1 NM_003779.3 XM_005245566.1 XM_011510093.1 XM_017002714.1 XM_017002715.1 XM_005245291.3 XM_005245295.3 XM_006711402.2 XM_006711403.2 XM_006711406.2 XM_011509663.2 XM_011509664.1 XM_011509665.2 XM_011509666.2 XM_011509667.2 XM_011509668.2 XM_011509669.1 XM_011509670.2 XM_011509673.2 XM_011509674.2 XM_011509676.1 XM_011509677.1 XM_011509678.1 XM_011509679.1 XM_011509681.1 XM_011509682.1 XM_017001561.1 XM_017001562.1 XM_017001563.1 XM_017001564.1 XM_017001570.1 XM_017001571.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
676662 c.583G>A p.D195N Chr.1: 161173956-161173956 on Build GRCh38
2086855 c.539G>A p.R180Q Chr.1: 161174000-161174000 on Build GRCh38
4776945 c.500G>A p.G167E Chr.1: 161174039-161174039 on Build GRCh38
6227720 c.550C>T p.R184C Chr.1: 161173989-161173989 on Build GRCh38
6202475 c.668A>G p.K223R Chr.1: 161173871-161173871 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751401924 Chr.1: 161173988-161173988 on Build GRCh38
rs763198781 Chr.1: 161173967-161173967 on Build GRCh38
rs756962425 Chr.1: 161174000-161174000 on Build GRCh38
rs201368066 Chr.1: 161174010-161174010 on Build GRCh38
rs918014408 Chr.1: 161173916-161173917 on Build GRCh38
rs3813619 Chr.1: 161174011-161174011 on Build GRCh38
rs373721033 Chr.1: 161173956-161173956 on Build GRCh38
rs748981503 Chr.1: 161173904-161173904 on Build GRCh38
rs991069779 Chr.1: 161173935-161173935 on Build GRCh38
rs1022664030 Chr.1: 161173946-161173946 on Build GRCh38
rs769132019 Chr.1: 161174056-161174056 on Build GRCh38
rs755642811 Chr.1: 161173885-161173885 on Build GRCh38
rs752663662 Chr.1: 161174003-161174003 on Build GRCh38
rs111476933 Chr.1: 161173858-161173858 on Build GRCh38
rs762759113 Chr.1: 161173851-161173851 on Build GRCh38
rs936644055 Chr.1: 161174055-161174055 on Build GRCh38
rs770641073 Chr.1: 161173907-161173907 on Build GRCh38
rs776027319 Chr.1: 161173908-161173908 on Build GRCh38
rs756992272 Chr.1: 161174041-161174041 on Build GRCh38
rs199582443 Chr.1: 161173963-161173963 on Build GRCh38
rs371653673 Chr.1: 161173966-161173966 on Build GRCh38
rs58449083 Chr.1: 161173947-161173947 on Build GRCh38
rs755946197 Chr.1: 161174009-161174009 on Build GRCh38
rs769328177 Chr.1: 161173925-161173925 on Build GRCh38
rs766543811 Chr.1: 161173974-161173974 on Build GRCh38
rs958312066 Chr.1: 161173909-161173909 on Build GRCh38
rs781081815 Chr.1: 161173851-161173851 on Build GRCh38
rs948091801 Chr.1: 161173931-161173931 on Build GRCh38
rs778678677 Chr.1: 161174048-161174048 on Build GRCh38
rs754732272 Chr.1: 161173992-161173992 on Build GRCh38
rs545295972 Chr.1: 161173884-161173884 on Build GRCh38
rs747356159 Chr.1: 161173921-161173921 on Build GRCh38
rs187580965 Chr.1: 161173957-161173957 on Build GRCh38
rs200391077 Chr.1: 161173896-161173896 on Build GRCh38
rs776713363 Chr.1: 161173905-161173905 on Build GRCh38
rs549057226 Chr.1: 161173923-161173923 on Build GRCh38
rs375281010 Chr.1: 161174052-161174052 on Build GRCh38
rs199539404 Chr.1: 161173895-161173895 on Build GRCh38

View Full Product Details