Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 193181202-193181445 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GALT2
Entrez Gene ID
8707
Gene Name
beta-1,3-galactosyltransferase 2
Gene Aliases
BETA3GALT2, GLCT2, beta3Gal-T2
UniGene
Hs.518834
Gene Chromosome Location
Chr.1: 193178730-193186613 on Build GRCh38
Transcripts
NM_003783.3 NM_024529.4

Gene Symbol
CDC73
Entrez Gene ID
79577
Gene Name
cell division cycle 73
Gene Aliases
C1orf28, FIHP, HPTJT, HRPT1, HRPT2, HYX
UniGene
Hs.378996
Gene Chromosome Location
Chr.1: 193121958-193254815 on Build GRCh38
Transcripts
NM_003783.3 NM_024529.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6215389 c.357T>G p.N119K Chr.1: 193181206-193181206 on Build GRCh38
5052885 c.349A>G p.S117G Chr.1: 193181214-193181214 on Build GRCh38
4026325 c.128T>A p.F43Y Chr.1: 193181435-193181435 on Build GRCh38
901396 c.238C>T p.R80W Chr.1: 193181325-193181325 on Build GRCh38
274110 c.202C>T p.R68W Chr.1: 193181361-193181361 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1053348927 Chr.1: 193181269-193181269 on Build GRCh38
rs761625282 Chr.1: 193181392-193181394 on Build GRCh38
rs768636884 Chr.1: 193181302-193181302 on Build GRCh38
rs780961822 Chr.1: 193181263-193181263 on Build GRCh38
rs147537643 Chr.1: 193181265-193181265 on Build GRCh38
rs759306050 Chr.1: 193181344-193181344 on Build GRCh38
rs770667754 Chr.1: 193181254-193181254 on Build GRCh38
rs751156224 Chr.1: 193181361-193181361 on Build GRCh38
rs746284962 Chr.1: 193181279-193181279 on Build GRCh38
rs201995399 Chr.1: 193181297-193181297 on Build GRCh38
rs758297188 Chr.1: 193181354-193181354 on Build GRCh38
rs182305763 Chr.1: 193181360-193181360 on Build GRCh38
rs780411189 Chr.1: 193181371-193181371 on Build GRCh38
rs774804871 Chr.1: 193181405-193181405 on Build GRCh38
rs752581247 Chr.1: 193181347-193181347 on Build GRCh38
rs576603378 Chr.1: 193181324-193181324 on Build GRCh38
rs767764174 Chr.1: 193181325-193181325 on Build GRCh38
rs746673079 Chr.1: 193181230-193181230 on Build GRCh38
rs774361824 Chr.1: 193181321-193181321 on Build GRCh38
rs946591897 Chr.1: 193181415-193181415 on Build GRCh38
rs776292714 Chr.1: 193181378-193181379 on Build GRCh38
rs969040425 Chr.1: 193181240-193181240 on Build GRCh38
rs138446152 Chr.1: 193181410-193181410 on Build GRCh38
rs769161946 Chr.1: 193181396-193181396 on Build GRCh38
rs781051676 Chr.1: 193181367-193181367 on Build GRCh38
rs770182939 Chr.1: 193181283-193181283 on Build GRCh38
rs750619305 Chr.1: 193181336-193181336 on Build GRCh38
rs200584199 Chr.1: 193181301-193181301 on Build GRCh38
rs745513357 Chr.1: 193181369-193181369 on Build GRCh38
rs765075342 Chr.1: 193181345-193181345 on Build GRCh38
rs369418702 Chr.1: 193181389-193181389 on Build GRCh38
rs775932344 Chr.1: 193181287-193181287 on Build GRCh38
rs372960322 Chr.1: 193181413-193181413 on Build GRCh38

View Full Product Details