Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 89209772-89210037 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHA3
Entrez Gene ID
2042
Gene Name
EPH receptor A3
Gene Aliases
EK4, ETK, ETK1, HEK, HEK4, TYRO4
UniGene
Hs.123642
Gene Chromosome Location
Chr.3: 89107524-89482134 on Build GRCh38
Transcripts
NM_005233.5 NM_182644.2 XM_005264715.2 XM_005264716.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6984833 c.160G>A p.E54K Chr.3: 89209866-89209866 on Build GRCh38
6165568 c.155G>A p.W52* Chr.3: 89209861-89209861 on Build GRCh38
6934137 c.313G>A p.D105N Chr.3: 89210019-89210019 on Build GRCh38
2828269 c.219C>G p.V73V Chr.3: 89209925-89209925 on Build GRCh38
6321082 c.155G>T p.W52L Chr.3: 89209861-89209861 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1019076961 Chr.3: 89209954-89209954 on Build GRCh38
rs778365543 Chr.3: 89209935-89209935 on Build GRCh38
rs760579355 Chr.3: 89210012-89210012 on Build GRCh38
rs371194762 Chr.3: 89209984-89209984 on Build GRCh38
rs1027305939 Chr.3: 89210003-89210003 on Build GRCh38
rs201901482 Chr.3: 89209854-89209854 on Build GRCh38
rs778273718 Chr.3: 89209809-89209809 on Build GRCh38
rs372915459 Chr.3: 89209819-89209819 on Build GRCh38
rs767507555 Chr.3: 89209835-89209835 on Build GRCh38
rs149992625 Chr.3: 89209874-89209874 on Build GRCh38
rs148063788 Chr.3: 89209850-89209850 on Build GRCh38
rs761779176 Chr.3: 89209830-89209830 on Build GRCh38
rs267599943 Chr.3: 89209929-89209929 on Build GRCh38
rs558029690 Chr.3: 89209997-89209997 on Build GRCh38
rs778392937 Chr.3: 89209921-89209921 on Build GRCh38
rs771292552 Chr.3: 89209968-89209968 on Build GRCh38
rs539750746 Chr.3: 89209981-89209981 on Build GRCh38
rs771486126 Chr.3: 89209880-89209880 on Build GRCh38
rs747296143 Chr.3: 89209952-89209952 on Build GRCh38
rs565897036 Chr.3: 89209933-89209933 on Build GRCh38
rs142484873 Chr.3: 89209972-89209972 on Build GRCh38
rs771868437 Chr.3: 89209995-89209995 on Build GRCh38
rs754685073 Chr.3: 89209877-89209877 on Build GRCh38
rs752206333 Chr.3: 89209931-89209931 on Build GRCh38
rs753507903 Chr.3: 89209876-89209876 on Build GRCh38
rs775767746 Chr.3: 89209829-89209829 on Build GRCh38
rs200290249 Chr.3: 89209991-89209991 on Build GRCh38
rs369150999 Chr.3: 89209988-89209988 on Build GRCh38
rs567227921 Chr.3: 89209810-89209810 on Build GRCh38

View Full Product Details