Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 69843078-69843321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMCL1
Entrez Gene ID
64395
Gene Name
germ cell-less, spermatogenesis associated 1
Gene Aliases
BTBD13, GCL, GCL1, SPATA29
UniGene
Hs.293971
Gene Chromosome Location
Chr.2: 69829606-69881396 on Build GRCh38
Transcripts
NM_178439.4 XM_011533033.1 XM_011533034.2 XM_017004704.1 XM_017004705.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1022459 c.582C>T p.D194D Chr.2: 69843151-69843151 on Build GRCh38
6435288 c.635C>A p.T212N Chr.2: 69843204-69843204 on Build GRCh38
5809866 c.637G>A p.V213I Chr.2: 69843206-69843206 on Build GRCh38
1022460 c.642T>C p.C214C Chr.2: 69843211-69843211 on Build GRCh38
5891663 c.584G>A p.G195D Chr.2: 69843153-69843153 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771942543 Chr.2: 69843110-69843110 on Build GRCh38
rs755175408 Chr.2: 69843244-69843244 on Build GRCh38
rs770860619 Chr.2: 69843114-69843114 on Build GRCh38
rs372890545 Chr.2: 69843269-69843269 on Build GRCh38
rs746874561 Chr.2: 69843260-69843260 on Build GRCh38
rs146826393 Chr.2: 69843188-69843188 on Build GRCh38
rs750948696 Chr.2: 69843209-69843209 on Build GRCh38
rs560427399 Chr.2: 69843213-69843213 on Build GRCh38
rs762549347 Chr.2: 69843166-69843166 on Build GRCh38
rs367702157 Chr.2: 69843208-69843208 on Build GRCh38
rs745775283 Chr.2: 69843288-69843288 on Build GRCh38
rs148776259 Chr.2: 69843151-69843151 on Build GRCh38
rs775252833 Chr.2: 69843158-69843158 on Build GRCh38
rs779965071 Chr.2: 69843221-69843221 on Build GRCh38
rs753896134 Chr.2: 69843111-69843111 on Build GRCh38
rs755400969 Chr.2: 69843252-69843252 on Build GRCh38
rs72895274 Chr.2: 69843259-69843259 on Build GRCh38
rs751988390 Chr.2: 69843184-69843184 on Build GRCh38
rs906577682 Chr.2: 69843264-69843264 on Build GRCh38
rs946321190 Chr.2: 69843121-69843121 on Build GRCh38
rs910927571 Chr.2: 69843116-69843116 on Build GRCh38
rs756103122 Chr.2: 69843241-69843241 on Build GRCh38
rs751838716 Chr.2: 69843228-69843229 on Build GRCh38
rs763756043 Chr.2: 69843177-69843177 on Build GRCh38
rs764631481 Chr.2: 69843152-69843152 on Build GRCh38
rs60400242 Chr.2: 69843287-69843287 on Build GRCh38
rs1000356699 Chr.2: 69843251-69843251 on Build GRCh38

View Full Product Details