Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 9967861-9968122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL5A3
Entrez Gene ID
50509
Gene Name
collagen type V alpha 3
Gene Aliases
-
UniGene
Hs.235368
Gene Chromosome Location
Chr.19: 9959561-10010532 on Build GRCh38
Transcripts
NM_015719.3 XM_011528042.2 XM_017026849.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7362248 c.4376T>C p.L1459P Chr.19: 9967932-9967932 on Build GRCh38
7098092 c.4343G>A p.G1448D Chr.19: 9968051-9968051 on Build GRCh38
7125576 c.4391C>T p.S1464L Chr.19: 9967917-9967917 on Build GRCh38
4131127 c.4395A>G p.K1465K Chr.19: 9967913-9967913 on Build GRCh38
6992048 c.4391C>G p.S1464* Chr.19: 9967917-9967917 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777372677 Chr.19: 9968099-9968099 on Build GRCh38
rs780688189 Chr.19: 9967991-9967991 on Build GRCh38
rs199774264 Chr.19: 9967993-9967993 on Build GRCh38
rs769990667 Chr.19: 9967909-9967909 on Build GRCh38
rs754110974 Chr.19: 9967931-9967931 on Build GRCh38
rs113489314 Chr.19: 9968096-9968096 on Build GRCh38
rs371587351 Chr.19: 9967920-9967920 on Build GRCh38
rs765408888 Chr.19: 9967963-9967963 on Build GRCh38
rs759036194 Chr.19: 9967985-9967985 on Build GRCh38
rs750583497 Chr.19: 9968071-9968071 on Build GRCh38
rs376148241 Chr.19: 9968062-9968062 on Build GRCh38
rs755824016 Chr.19: 9968098-9968098 on Build GRCh38
rs762881841 Chr.19: 9967898-9967898 on Build GRCh38
rs147777886 Chr.19: 9968093-9968093 on Build GRCh38
rs374717602 Chr.19: 9968018-9968018 on Build GRCh38
rs3815748 Chr.19: 9967913-9967913 on Build GRCh38
rs761880871 Chr.19: 9967937-9967937 on Build GRCh38
rs769369864 Chr.19: 9968017-9968017 on Build GRCh38
rs766529278 Chr.19: 9968091-9968091 on Build GRCh38
rs560163996 Chr.19: 9967984-9967984 on Build GRCh38
rs774300689 Chr.19: 9968040-9968040 on Build GRCh38
rs8109497 Chr.19: 9968095-9968095 on Build GRCh38
rs765540234 Chr.19: 9968069-9968069 on Build GRCh38
rs752132424 Chr.19: 9968092-9968092 on Build GRCh38
rs370250990 Chr.19: 9967881-9967881 on Build GRCh38
rs748900003 Chr.19: 9968100-9968100 on Build GRCh38
rs368508428 Chr.19: 9968059-9968059 on Build GRCh38
rs770852092 Chr.19: 9967904-9967904 on Build GRCh38
rs747852601 Chr.19: 9967998-9967998 on Build GRCh38
rs759341052 Chr.19: 9968041-9968041 on Build GRCh38
rs746675031 Chr.19: 9967985-9967986 on Build GRCh38
rs763099096 Chr.19: 9968080-9968080 on Build GRCh38
rs533616651 Chr.19: 9968039-9968039 on Build GRCh38
rs199586807 Chr.19: 9968027-9968027 on Build GRCh38
rs777331845 Chr.19: 9968026-9968026 on Build GRCh38
rs775077814 Chr.19: 9968064-9968064 on Build GRCh38
rs755405036 Chr.19: 9967994-9967994 on Build GRCh38
rs762158108 Chr.19: 9968067-9968067 on Build GRCh38
rs538640420 Chr.19: 9967984-9967984 on Build GRCh38
rs571945809 Chr.19: 9967905-9967905 on Build GRCh38
rs373652963 Chr.19: 9967884-9967884 on Build GRCh38
rs750522305 Chr.19: 9967969-9967969 on Build GRCh38
rs76653582 Chr.19: 9968022-9968022 on Build GRCh38
rs368019895 Chr.19: 9967885-9967885 on Build GRCh38

View Full Product Details