Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 203179532-203179769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHI3L1
Entrez Gene ID
1116
Gene Name
chitinase 3 like 1
Gene Aliases
ASRT7, CGP-39, GP-39, GP39, HC-gp39, HCGP-3P, YKL-40, YKL40, YYL-40, hCGP-39
UniGene
Hs.382202
Gene Chromosome Location
Chr.1: 203178931-203186794 on Build GRCh38
Transcripts
NM_001276.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4167683 c.1011G>A p.K337K Chr.1: 203179761-203179761 on Build GRCh38
6508502 c.1011G>T p.K337N Chr.1: 203179761-203179761 on Build GRCh38
1337973 c.1039G>A p.A347T Chr.1: 203179558-203179558 on Build GRCh38
2213568 c.1006A>C p.S336R Chr.1: 203179766-203179766 on Build GRCh38
902451 c.1040C>T p.A347V Chr.1: 203179557-203179557 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs374657390 Chr.1: 203179709-203179709 on Build GRCh38
rs141727518 Chr.1: 203179740-203179740 on Build GRCh38
rs1020411445 Chr.1: 203179682-203179682 on Build GRCh38
rs201199973 Chr.1: 203179725-203179725 on Build GRCh38
rs114984905 Chr.1: 203179570-203179570 on Build GRCh38
rs746542665 Chr.1: 203179661-203179661 on Build GRCh38
rs756157745 Chr.1: 203179708-203179708 on Build GRCh38
rs371798594 Chr.1: 203179745-203179745 on Build GRCh38
rs371109734 Chr.1: 203179622-203179622 on Build GRCh38
rs929786186 Chr.1: 203179571-203179571 on Build GRCh38
rs1130572 Chr.1: 203179568-203179568 on Build GRCh38
rs759287286 Chr.1: 203179674-203179674 on Build GRCh38
rs762740927 Chr.1: 203179689-203179689 on Build GRCh38
rs758785010 Chr.1: 203179739-203179739 on Build GRCh38
rs917495936 Chr.1: 203179741-203179741 on Build GRCh38
rs748598425 Chr.1: 203179644-203179644 on Build GRCh38
rs758424133 Chr.1: 203179604-203179604 on Build GRCh38
rs753285691 Chr.1: 203179711-203179711 on Build GRCh38
rs749554697 Chr.1: 203179625-203179625 on Build GRCh38
rs958993622 Chr.1: 203179735-203179735 on Build GRCh38
rs376352545 Chr.1: 203179554-203179554 on Build GRCh38
rs776398402 Chr.1: 203179668-203179668 on Build GRCh38
rs549780563 Chr.1: 203179710-203179710 on Build GRCh38
rs764113488 Chr.1: 203179691-203179691 on Build GRCh38
rs756278618 Chr.1: 203179617-203179617 on Build GRCh38
rs567485587 Chr.1: 203179679-203179679 on Build GRCh38
rs773378246 Chr.1: 203179718-203179720 on Build GRCh38
rs367660199 Chr.1: 203179608-203179608 on Build GRCh38
rs769580170 Chr.1: 203179710-203179710 on Build GRCh38
rs779084360 Chr.1: 203179736-203179736 on Build GRCh38
rs755277452 Chr.1: 203179630-203179630 on Build GRCh38
rs772643074 Chr.1: 203179646-203179646 on Build GRCh38
rs753123504 Chr.1: 203179738-203179738 on Build GRCh38
rs779392799 Chr.1: 203179640-203179640 on Build GRCh38
rs750462113 Chr.1: 203179698-203179698 on Build GRCh38
rs112567571 Chr.1: 203179552-203179552 on Build GRCh38
rs370477111 Chr.1: 203179572-203179572 on Build GRCh38
rs146151706 Chr.1: 203179553-203179553 on Build GRCh38
rs984368931 Chr.1: 203179579-203179579 on Build GRCh38
rs755294971 Chr.1: 203179727-203179727 on Build GRCh38
rs770669922 Chr.1: 203179663-203179663 on Build GRCh38
rs772794137 Chr.1: 203179650-203179650 on Build GRCh38
rs775359723 Chr.1: 203179688-203179688 on Build GRCh38
rs750514890 Chr.1: 203179615-203179615 on Build GRCh38
rs1049410 Chr.1: 203179556-203179556 on Build GRCh38
rs780284642 Chr.1: 203179621-203179621 on Build GRCh38
rs373596867 Chr.1: 203179599-203179599 on Build GRCh38
rs527531482 Chr.1: 203179557-203179557 on Build GRCh38
rs947820557 Chr.1: 203179616-203179616 on Build GRCh38

View Full Product Details