Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 23764506-23764763 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C22orf15
Entrez Gene ID
150248
Gene Name
chromosome 22 open reading frame 15
Gene Aliases
N27C7-3
UniGene
Hs.116254
Gene Chromosome Location
Chr.22: 23762522-23765863 on Build GRCh38
Transcripts
NM_182520.2 XM_011529907.2 XM_011529908.2 XM_011529912.2 XM_017028602.1 XM_017028603.1 XM_017028604.1 XM_017028605.1 XM_017028606.1 XM_017028607.1 XM_017028608.1 XM_017028609.1 XM_017028610.1 XM_017028611.1 XM_017028612.1 XM_017028613.1 XM_017028614.1 XM_017028615.1 XM_017028616.1 XM_017028617.1 XM_017028618.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5432158 c.173G>A p.S58N Chr.22: 23764706-23764706 on Build GRCh38
5527710 c.251-1G>A p.? Chr.22: 23764638-23764638 on Build GRCh38
5393244 c.326A>C p.Y109S Chr.22: 23764699-23764699 on Build GRCh38
5527711 c.105G>A p.Q35Q Chr.22: 23764638-23764638 on Build GRCh38
5432159 c.333G>A p.E111E Chr.22: 23764706-23764706 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs543669757 Chr.22: 23764685-23764685 on Build GRCh38
rs554903635 Chr.22: 23764653-23764653 on Build GRCh38
rs758186435 Chr.22: 23764713-23764713 on Build GRCh38
rs749377402 Chr.22: 23764743-23764743 on Build GRCh38
rs566868267 Chr.22: 23764640-23764640 on Build GRCh38
rs781298027 Chr.22: 23764613-23764615 on Build GRCh38
rs776435213 Chr.22: 23764690-23764690 on Build GRCh38
rs751455317 Chr.22: 23764726-23764726 on Build GRCh38
rs569317011 Chr.22: 23764587-23764587 on Build GRCh38
rs751076431 Chr.22: 23764558-23764559 on Build GRCh38
rs554034284 Chr.22: 23764683-23764683 on Build GRCh38
rs759166916 Chr.22: 23764694-23764694 on Build GRCh38
rs775663786 Chr.22: 23764560-23764560 on Build GRCh38
rs764038416 Chr.22: 23764725-23764725 on Build GRCh38
rs746415942 Chr.22: 23764641-23764642 on Build GRCh38
rs145039987 Chr.22: 23764619-23764619 on Build GRCh38
rs758333362 Chr.22: 23764633-23764633 on Build GRCh38
rs763845261 Chr.22: 23764528-23764528 on Build GRCh38
rs752475063 Chr.22: 23764710-23764710 on Build GRCh38
rs752522586 Chr.22: 23764630-23764630 on Build GRCh38
rs148288019 Chr.22: 23764577-23764577 on Build GRCh38
rs969563923 Chr.22: 23764530-23764530 on Build GRCh38
rs112223389 Chr.22: 23764554-23764554 on Build GRCh38
rs964907974 Chr.22: 23764613-23764613 on Build GRCh38
rs763394298 Chr.22: 23764561-23764561 on Build GRCh38
rs147550502 Chr.22: 23764644-23764644 on Build GRCh38
rs756188575 Chr.22: 23764732-23764732 on Build GRCh38
rs756496280 Chr.22: 23764732-23764732 on Build GRCh38
rs376388637 Chr.22: 23764662-23764662 on Build GRCh38
rs764937850 Chr.22: 23764697-23764697 on Build GRCh38
rs1009156697 Chr.22: 23764585-23764585 on Build GRCh38
rs768859927 Chr.22: 23764572-23764572 on Build GRCh38
rs534046925 Chr.22: 23764676-23764676 on Build GRCh38
rs774929772 Chr.22: 23764573-23764573 on Build GRCh38
rs756851140 Chr.22: 23764609-23764622 on Build GRCh38
rs145628774 Chr.22: 23764672-23764672 on Build GRCh38
rs140216229 Chr.22: 23764663-23764663 on Build GRCh38
rs770038408 Chr.22: 23764535-23764535 on Build GRCh38
rs546921891 Chr.22: 23764624-23764624 on Build GRCh38
rs774640235 Chr.22: 23764665-23764665 on Build GRCh38
rs760231662 Chr.22: 23764600-23764600 on Build GRCh38
rs773833552 Chr.22: 23764682-23764682 on Build GRCh38
rs776068105 Chr.22: 23764736-23764736 on Build GRCh38
rs780053397 Chr.22: 23764738-23764738 on Build GRCh38
rs754835786 Chr.22: 23764620-23764620 on Build GRCh38

View Full Product Details