Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129257659-129257918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPG1
Entrez Gene ID
22820
Gene Name
coatomer protein complex subunit gamma 1
Gene Aliases
COPG
UniGene
Hs.518250
Gene Chromosome Location
Chr.3: 129249575-129277773 on Build GRCh38
Transcripts
NM_016128.3 XM_011512549.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5915132 c.817G>A p.A273T Chr.3: 129257806-129257806 on Build GRCh38
4435263 c.859C>A p.P287T Chr.3: 129257848-129257848 on Build GRCh38
5549391 c.864T>C p.A288A Chr.3: 129257853-129257853 on Build GRCh38
4318820 c.778C>T p.R260C Chr.3: 129257767-129257767 on Build GRCh38
5863457 c.811G>A p.A271T Chr.3: 129257800-129257800 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776259331 Chr.3: 129257722-129257722 on Build GRCh38
rs748661124 Chr.3: 129257809-129257809 on Build GRCh38
rs200701430 Chr.3: 129257791-129257791 on Build GRCh38
rs147163571 Chr.3: 129257799-129257799 on Build GRCh38
rs143558091 Chr.3: 129257729-129257729 on Build GRCh38
rs370430335 Chr.3: 129257691-129257691 on Build GRCh38
rs760787011 Chr.3: 129257819-129257819 on Build GRCh38
rs200402729 Chr.3: 129257692-129257692 on Build GRCh38
rs758465944 Chr.3: 129257849-129257849 on Build GRCh38
rs4927952 Chr.3: 129257820-129257820 on Build GRCh38
rs766357893 Chr.3: 129257754-129257754 on Build GRCh38
rs4927951 Chr.3: 129257897-129257897 on Build GRCh38
rs374144434 Chr.3: 129257865-129257865 on Build GRCh38
rs377596678 Chr.3: 129257778-129257778 on Build GRCh38
rs371600636 Chr.3: 129257876-129257876 on Build GRCh38
rs114007375 Chr.3: 129257886-129257886 on Build GRCh38
rs764256530 Chr.3: 129257866-129257867 on Build GRCh38
rs748642487 Chr.3: 129257884-129257884 on Build GRCh38
rs201593901 Chr.3: 129257769-129257769 on Build GRCh38
rs759602147 Chr.3: 129257836-129257836 on Build GRCh38
rs370226896 Chr.3: 129257800-129257800 on Build GRCh38
rs77129536 Chr.3: 129257850-129257850 on Build GRCh38
rs773358694 Chr.3: 129257745-129257745 on Build GRCh38
rs765410626 Chr.3: 129257838-129257838 on Build GRCh38
rs753715514 Chr.3: 129257690-129257690 on Build GRCh38
rs772423585 Chr.3: 129257811-129257811 on Build GRCh38
rs751406592 Chr.3: 129257851-129257851 on Build GRCh38
rs1004991354 Chr.3: 129257844-129257844 on Build GRCh38
rs375319623 Chr.3: 129257755-129257755 on Build GRCh38
rs767885335 Chr.3: 129257735-129257735 on Build GRCh38
rs145564919 Chr.3: 129257697-129257697 on Build GRCh38
rs769213450 Chr.3: 129257805-129257805 on Build GRCh38
rs755556647 Chr.3: 129257875-129257875 on Build GRCh38
rs780852410 Chr.3: 129257854-129257854 on Build GRCh38
rs933444140 Chr.3: 129257796-129257796 on Build GRCh38
rs752693406 Chr.3: 129257779-129257779 on Build GRCh38
rs772617478 Chr.3: 129257885-129257885 on Build GRCh38
rs1035368863 Chr.3: 129257869-129257869 on Build GRCh38
rs995344366 Chr.3: 129257765-129257765 on Build GRCh38
rs755044271 Chr.3: 129257772-129257772 on Build GRCh38
rs753905985 Chr.3: 129257891-129257892 on Build GRCh38
rs375030618 Chr.3: 129257804-129257804 on Build GRCh38
rs778550950 Chr.3: 129257696-129257696 on Build GRCh38
rs746662896 Chr.3: 129257712-129257712 on Build GRCh38
rs761026368 Chr.3: 129257751-129257751 on Build GRCh38
rs777508581 Chr.3: 129257708-129257708 on Build GRCh38
rs776740895 Chr.3: 129257823-129257823 on Build GRCh38
rs763343530 Chr.3: 129257723-129257723 on Build GRCh38
rs780188286 Chr.3: 129257688-129257688 on Build GRCh38
rs199570967 Chr.3: 129257702-129257702 on Build GRCh38
rs770341512 Chr.3: 129257718-129257718 on Build GRCh38
rs769241698 Chr.3: 129257727-129257727 on Build GRCh38
rs752675074 Chr.3: 129257846-129257846 on Build GRCh38
rs774874304 Chr.3: 129257728-129257728 on Build GRCh38
rs745524558 Chr.3: 129257874-129257874 on Build GRCh38
rs368509570 Chr.3: 129257816-129257816 on Build GRCh38
rs758197972 Chr.3: 129257781-129257781 on Build GRCh38
rs923440210 Chr.3: 129257785-129257785 on Build GRCh38

View Full Product Details