Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 43492737-43492978 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf34
Entrez Gene ID
375444
Gene Name
chromosome 5 open reading frame 34
Gene Aliases
-
UniGene
Hs.661641
Gene Chromosome Location
Chr.5: 43486699-43515433 on Build GRCh38
Transcripts
NM_198566.3 XM_011514038.2 XM_017009445.1 XM_017009446.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6253793 c.1448T>C p.L483P Chr.5: 43492757-43492757 on Build GRCh38
6251244 c.1455G>A p.W485* Chr.5: 43492750-43492750 on Build GRCh38
4381599 c.1325T>C p.I442T Chr.5: 43492880-43492880 on Build GRCh38
1068432 c.1390C>A p.L464I Chr.5: 43492815-43492815 on Build GRCh38
3828113 c.1394C>A p.A465D Chr.5: 43492811-43492811 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779102760 Chr.5: 43492882-43492882 on Build GRCh38
rs756388677 Chr.5: 43492868-43492868 on Build GRCh38
rs150186169 Chr.5: 43492912-43492912 on Build GRCh38
rs759309101 Chr.5: 43492803-43492803 on Build GRCh38
rs374353571 Chr.5: 43492849-43492849 on Build GRCh38
rs10039298 Chr.5: 43492876-43492876 on Build GRCh38
rs751282337 Chr.5: 43492938-43492938 on Build GRCh38
rs962564266 Chr.5: 43492789-43492789 on Build GRCh38
rs781373365 Chr.5: 43492852-43492852 on Build GRCh38
rs774930219 Chr.5: 43492790-43492790 on Build GRCh38
rs762622319 Chr.5: 43492839-43492839 on Build GRCh38
rs1024326512 Chr.5: 43492769-43492769 on Build GRCh38
rs141842516 Chr.5: 43492839-43492839 on Build GRCh38
rs769113086 Chr.5: 43492786-43492786 on Build GRCh38
rs187171845 Chr.5: 43492776-43492776 on Build GRCh38
rs377598879 Chr.5: 43492796-43492796 on Build GRCh38
rs772432252 Chr.5: 43492887-43492887 on Build GRCh38
rs775293435 Chr.5: 43492922-43492922 on Build GRCh38
rs549798238 Chr.5: 43492824-43492824 on Build GRCh38
rs780252251 Chr.5: 43492871-43492871 on Build GRCh38
rs370589007 Chr.5: 43492809-43492809 on Build GRCh38
rs997662994 Chr.5: 43492941-43492941 on Build GRCh38
rs751537516 Chr.5: 43492838-43492838 on Build GRCh38
rs769974753 Chr.5: 43492774-43492774 on Build GRCh38
rs559576933 Chr.5: 43492863-43492863 on Build GRCh38
rs762463532 Chr.5: 43492923-43492923 on Build GRCh38
rs762173212 Chr.5: 43492794-43492794 on Build GRCh38
rs569511344 Chr.5: 43492825-43492825 on Build GRCh38
rs145766724 Chr.5: 43492823-43492823 on Build GRCh38
rs765911966 Chr.5: 43492881-43492881 on Build GRCh38
rs776336155 Chr.5: 43492897-43492897 on Build GRCh38
rs763979938 Chr.5: 43492932-43492932 on Build GRCh38
rs748725763 Chr.5: 43492885-43492885 on Build GRCh38
rs769082135 Chr.5: 43492878-43492878 on Build GRCh38
rs769361001 Chr.5: 43492915-43492915 on Build GRCh38

View Full Product Details