Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 157285303-157285576 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYFIP2
Entrez Gene ID
26999
Gene Name
cytoplasmic FMR1 interacting protein 2
Gene Aliases
PIR121
UniGene
Hs.519702
Gene Chromosome Location
Chr.5: 157266080-157395598 on Build GRCh38
Transcripts
NM_001037333.2 NM_001291721.1 NM_001291722.1 NM_014376.3 XM_011534516.2 XM_017009341.1 XM_017009342.1

SNP Detail

SNP ID Chromosome Location
rs879875568 Chr.5: 157285406-157285406 on Build GRCh38
rs755029730 Chr.5: 157285409-157285409 on Build GRCh38
rs777945352 Chr.5: 157285433-157285433 on Build GRCh38
rs760411094 Chr.5: 157285360-157285360 on Build GRCh38
rs376687042 Chr.5: 157285400-157285400 on Build GRCh38
rs916715778 Chr.5: 157285543-157285543 on Build GRCh38
rs968323449 Chr.5: 157285545-157285545 on Build GRCh38
rs571089246 Chr.5: 157285528-157285528 on Build GRCh38
rs369640959 Chr.5: 157285472-157285472 on Build GRCh38
rs750421212 Chr.5: 157285403-157285403 on Build GRCh38
rs1011416477 Chr.5: 157285338-157285338 on Build GRCh38
rs757626231 Chr.5: 157285463-157285463 on Build GRCh38
rs761596947 Chr.5: 157285391-157285391 on Build GRCh38
rs78461959 Chr.5: 157285540-157285540 on Build GRCh38
rs767625240 Chr.5: 157285412-157285412 on Build GRCh38
rs371578292 Chr.5: 157285370-157285370 on Build GRCh38
rs775257977 Chr.5: 157285349-157285349 on Build GRCh38
rs1021631734 Chr.5: 157285341-157285341 on Build GRCh38
rs776506102 Chr.5: 157285373-157285373 on Build GRCh38
rs752822676 Chr.5: 157285430-157285430 on Build GRCh38
rs371710290 Chr.5: 157285470-157285470 on Build GRCh38
rs894433345 Chr.5: 157285331-157285331 on Build GRCh38
rs367998009 Chr.5: 157285442-157285442 on Build GRCh38
rs921423992 Chr.5: 157285482-157285482 on Build GRCh38
rs151110759 Chr.5: 157285535-157285535 on Build GRCh38
rs779546387 Chr.5: 157285508-157285508 on Build GRCh38

View Full Product Details