Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 68070671-68070922 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHKA
Entrez Gene ID
1119
Gene Name
choline kinase alpha
Gene Aliases
CHK, CK, CKI, EK
UniGene
Hs.77221
Gene Chromosome Location
Chr.11: 68052859-68121391 on Build GRCh38
Transcripts
NM_001277.2 NM_212469.1 XM_017017147.1 XM_017017148.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6426885 c.721C>A p.P241T Chr.11: 68070767-68070767 on Build GRCh38
931195 c.705T>G p.F235L Chr.11: 68070783-68070783 on Build GRCh38
1704305 c.647C>T p.T216I Chr.11: 68070841-68070841 on Build GRCh38
4801937 c.663G>A p.L221L Chr.11: 68070825-68070825 on Build GRCh38
931194 c.740A>C p.K247T Chr.11: 68070748-68070748 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775068715 Chr.11: 68070744-68070744 on Build GRCh38
rs17853641 Chr.11: 68070830-68070830 on Build GRCh38
rs752926601 Chr.11: 68070793-68070793 on Build GRCh38
rs374838678 Chr.11: 68070804-68070804 on Build GRCh38
rs370436517 Chr.11: 68070760-68070760 on Build GRCh38
rs767350207 Chr.11: 68070863-68070863 on Build GRCh38
rs552858667 Chr.11: 68070752-68070752 on Build GRCh38
rs775017205 Chr.11: 68070845-68070845 on Build GRCh38
rs768349446 Chr.11: 68070850-68070850 on Build GRCh38
rs143616553 Chr.11: 68070849-68070849 on Build GRCh38
rs374700961 Chr.11: 68070872-68070872 on Build GRCh38
rs375282627 Chr.11: 68070712-68070712 on Build GRCh38
rs935864448 Chr.11: 68070759-68070759 on Build GRCh38
rs201285000 Chr.11: 68070801-68070801 on Build GRCh38
rs761788836 Chr.11: 68070772-68070772 on Build GRCh38
rs367870361 Chr.11: 68070750-68070750 on Build GRCh38
rs753948663 Chr.11: 68070787-68070787 on Build GRCh38
rs751354041 Chr.11: 68070770-68070770 on Build GRCh38
rs760724394 Chr.11: 68070874-68070874 on Build GRCh38
rs765376469 Chr.11: 68070889-68070889 on Build GRCh38
rs368348971 Chr.11: 68070883-68070883 on Build GRCh38
rs368758229 Chr.11: 68070827-68070827 on Build GRCh38
rs769249113 Chr.11: 68070836-68070836 on Build GRCh38
rs149289548 Chr.11: 68070822-68070822 on Build GRCh38
rs368477871 Chr.11: 68070853-68070853 on Build GRCh38
rs945709300 Chr.11: 68070816-68070816 on Build GRCh38
rs756627931 Chr.11: 68070768-68070768 on Build GRCh38
rs759031586 Chr.11: 68070711-68070711 on Build GRCh38
rs370716716 Chr.11: 68070695-68070695 on Build GRCh38
rs138597091 Chr.11: 68070800-68070800 on Build GRCh38
rs763231816 Chr.11: 68070698-68070698 on Build GRCh38
rs773743414 Chr.11: 68070696-68070696 on Build GRCh38
rs888867688 Chr.11: 68070779-68070779 on Build GRCh38
rs779155040 Chr.11: 68070789-68070789 on Build GRCh38
rs771462394 Chr.11: 68070812-68070812 on Build GRCh38
rs747517911 Chr.11: 68070699-68070699 on Build GRCh38
rs527698957 Chr.11: 68070703-68070703 on Build GRCh38
rs767414264 Chr.11: 68070778-68070778 on Build GRCh38
rs761582548 Chr.11: 68070854-68070854 on Build GRCh38

View Full Product Details