Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 36142473-36142710 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPNS1
Entrez Gene ID
826
Gene Name
calpain small subunit 1
Gene Aliases
CALPAIN4, CANP, CANPS, CAPN4, CDPS, CSS1
UniGene
Hs.515371
Gene Chromosome Location
Chr.19: 36139926-36150353 on Build GRCh38
Transcripts
NM_001003962.2 NM_001302632.1 NM_001302633.1 NM_001749.3 XM_005259295.1 XM_005259296.1 XM_011527359.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2150428 c.292G>T p.V98F Chr.19: 36142700-36142700 on Build GRCh38
5028552 c.282G>C p.E94D Chr.19: 36142690-36142690 on Build GRCh38
3959969 c.278A>G p.N93S Chr.19: 36142686-36142686 on Build GRCh38
1393051 c.292G>A p.V98I Chr.19: 36142700-36142700 on Build GRCh38
6619950 c.261C>T p.Y87Y Chr.19: 36142669-36142669 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374159469 Chr.19: 36142596-36142596 on Build GRCh38
rs777534419 Chr.19: 36142651-36142651 on Build GRCh38
rs113501827 Chr.19: 36142650-36142650 on Build GRCh38
rs766391056 Chr.19: 36142548-36142548 on Build GRCh38
rs202081010 Chr.19: 36142671-36142671 on Build GRCh38
rs757940775 Chr.19: 36142646-36142646 on Build GRCh38
rs373217382 Chr.19: 36142645-36142645 on Build GRCh38
rs755984538 Chr.19: 36142627-36142627 on Build GRCh38
rs1010089622 Chr.19: 36142558-36142558 on Build GRCh38
rs767610570 Chr.19: 36142682-36142682 on Build GRCh38
rs761953872 Chr.19: 36142618-36142618 on Build GRCh38
rs866083785 Chr.19: 36142659-36142659 on Build GRCh38
rs371312155 Chr.19: 36142658-36142658 on Build GRCh38
rs1004197360 Chr.19: 36142515-36142515 on Build GRCh38
rs972968845 Chr.19: 36142692-36142692 on Build GRCh38
rs555333474 Chr.19: 36142509-36142509 on Build GRCh38
rs751647878 Chr.19: 36142617-36142617 on Build GRCh38
rs769151310 Chr.19: 36142670-36142670 on Build GRCh38
rs761900601 Chr.19: 36142677-36142677 on Build GRCh38
rs17878728 Chr.19: 36142532-36142532 on Build GRCh38
rs116114216 Chr.19: 36142657-36142657 on Build GRCh38
rs773523619 Chr.19: 36142683-36142683 on Build GRCh38
rs778499068 Chr.19: 36142643-36142643 on Build GRCh38
rs1001134137 Chr.19: 36142603-36142603 on Build GRCh38
rs753892238 Chr.19: 36142635-36142635 on Build GRCh38
rs747694630 Chr.19: 36142644-36142644 on Build GRCh38
rs770182217 Chr.19: 36142601-36142601 on Build GRCh38
rs776109817 Chr.19: 36142605-36142605 on Build GRCh38
rs750134961 Chr.19: 36142624-36142624 on Build GRCh38
rs560846443 Chr.19: 36142668-36142668 on Build GRCh38
rs559995874 Chr.19: 36142574-36142574 on Build GRCh38
rs766140029 Chr.19: 36142687-36142687 on Build GRCh38
rs779823964 Chr.19: 36142628-36142628 on Build GRCh38
rs113039946 Chr.19: 36142586-36142586 on Build GRCh38
rs763020514 Chr.19: 36142607-36142607 on Build GRCh38
rs961562994 Chr.19: 36142678-36142678 on Build GRCh38
rs367819061 Chr.19: 36142612-36142612 on Build GRCh38
rs760381510 Chr.19: 36142684-36142684 on Build GRCh38
rs767808433 Chr.19: 36142619-36142619 on Build GRCh38
rs754720342 Chr.19: 36142674-36142674 on Build GRCh38
rs775851016 Chr.19: 36142662-36142662 on Build GRCh38
rs1042981561 Chr.19: 36142582-36142582 on Build GRCh38

View Full Product Details