Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49808600-49808855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUZ
Entrez Gene ID
80199
Gene Name
fuzzy planar cell polarity protein
Gene Aliases
FY, NTD
UniGene
Hs.288800
Gene Chromosome Location
Chr.19: 49806866-49817376 on Build GRCh38
Transcripts
NM_001171937.1 NM_025129.4 NR_033269.1 XM_006723399.3 XM_011527339.1 XM_011527340.1 XM_011527341.2 XM_011527342.1 XM_011527343.1 XM_011527345.1 XM_011527346.1 XM_011527347.1 XM_017027319.1 XM_017027320.1 XM_017027321.1 XM_017027322.1 XM_017027323.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1304927 c.918G>A p.L306L Chr.19: 49808614-49808614 on Build GRCh38
5809563 c.922C>T p.R308C Chr.19: 49808610-49808610 on Build GRCh38
5973084 c.851G>T p.R284L Chr.19: 49808759-49808759 on Build GRCh38
6038242 c.796C>G p.R266G Chr.19: 49808814-49808814 on Build GRCh38
999441 c.901C>T p.L301F Chr.19: 49808631-49808631 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs368021335 Chr.19: 49808688-49808688 on Build GRCh38
rs919628292 Chr.19: 49808728-49808729 on Build GRCh38
rs751231939 Chr.19: 49808666-49808666 on Build GRCh38
rs757614328 Chr.19: 49808736-49808736 on Build GRCh38
rs548411113 Chr.19: 49808782-49808782 on Build GRCh38
rs775571739 Chr.19: 49808699-49808699 on Build GRCh38
rs772801618 Chr.19: 49808811-49808812 on Build GRCh38
rs752373370 Chr.19: 49808814-49808814 on Build GRCh38
rs374559980 Chr.19: 49808653-49808653 on Build GRCh38
rs867250115 Chr.19: 49808772-49808772 on Build GRCh38
rs765935647 Chr.19: 49808721-49808721 on Build GRCh38
rs913465891 Chr.19: 49808815-49808815 on Build GRCh38
rs201118677 Chr.19: 49808740-49808740 on Build GRCh38
rs771008942 Chr.19: 49808788-49808788 on Build GRCh38
rs925677932 Chr.19: 49808663-49808663 on Build GRCh38
rs747193091 Chr.19: 49808702-49808702 on Build GRCh38
rs746338643 Chr.19: 49808742-49808742 on Build GRCh38
rs11557714 Chr.19: 49808791-49808791 on Build GRCh38
rs757918578 Chr.19: 49808644-49808644 on Build GRCh38
rs141711433 Chr.19: 49808696-49808696 on Build GRCh38
rs993111578 Chr.19: 49808793-49808793 on Build GRCh38
rs542500278 Chr.19: 49808620-49808620 on Build GRCh38
rs141678080 Chr.19: 49808622-49808622 on Build GRCh38
rs751143762 Chr.19: 49808722-49808722 on Build GRCh38
rs143722232 Chr.19: 49808624-49808624 on Build GRCh38
rs775380683 Chr.19: 49808638-49808638 on Build GRCh38
rs746403012 Chr.19: 49808690-49808690 on Build GRCh38
rs201371956 Chr.19: 49808720-49808720 on Build GRCh38
rs763829848 Chr.19: 49808631-49808631 on Build GRCh38
rs766980819 Chr.19: 49808798-49808798 on Build GRCh38
rs780040869 Chr.19: 49808745-49808745 on Build GRCh38
rs758936919 Chr.19: 49808744-49808744 on Build GRCh38
rs548889142 Chr.19: 49808789-49808789 on Build GRCh38
rs900802753 Chr.19: 49808694-49808694 on Build GRCh38
rs368721486 Chr.19: 49808759-49808759 on Build GRCh38
rs749679018 Chr.19: 49808682-49808682 on Build GRCh38
rs776088329 Chr.19: 49808685-49808686 on Build GRCh38
rs532604554 Chr.19: 49808689-49808689 on Build GRCh38
rs781471549 Chr.19: 49808769-49808769 on Build GRCh38
rs200068950 Chr.19: 49808672-49808672 on Build GRCh38
rs776229517 Chr.19: 49808707-49808707 on Build GRCh38
rs762940657 Chr.19: 49808786-49808786 on Build GRCh38
rs747103238 Chr.19: 49808756-49808756 on Build GRCh38
rs754696650 Chr.19: 49808671-49808671 on Build GRCh38
rs763438505 Chr.19: 49808748-49808749 on Build GRCh38
rs769087470 Chr.19: 49808809-49808810 on Build GRCh38
rs765007508 Chr.19: 49808718-49808718 on Build GRCh38
rs760268753 Chr.19: 49808834-49808834 on Build GRCh38
rs370042440 Chr.19: 49808643-49808643 on Build GRCh38
rs1005328935 Chr.19: 49808731-49808731 on Build GRCh38
rs754622925 Chr.19: 49808723-49808723 on Build GRCh38
rs767188774 Chr.19: 49808725-49808725 on Build GRCh38
rs772901977 Chr.19: 49808784-49808784 on Build GRCh38
rs761655684 Chr.19: 49808708-49808708 on Build GRCh38
rs374092791 Chr.19: 49808632-49808632 on Build GRCh38
rs1001985054 Chr.19: 49808770-49808770 on Build GRCh38
rs1047400859 Chr.19: 49808701-49808701 on Build GRCh38
rs375035212 Chr.19: 49808700-49808700 on Build GRCh38
rs942375063 Chr.19: 49808667-49808667 on Build GRCh38
rs752362673 Chr.19: 49808735-49808735 on Build GRCh38
rs769605514 Chr.19: 49808783-49808783 on Build GRCh38
rs147900073 Chr.19: 49808621-49808621 on Build GRCh38

View Full Product Details