Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 36223134-36223401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOPEY2
Entrez Gene ID
9980
Gene Name
dopey family member 2
Gene Aliases
21orf5, C21orf5
UniGene
Hs.204575
Gene Chromosome Location
Chr.21: 36156781-36294274 on Build GRCh38
Transcripts
NM_001320714.1 NM_005128.3 XM_017028509.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1717272 c.1306A>G p.I436V Chr.21: 36223286-36223286 on Build GRCh38
4582063 c.1356T>C p.F452F Chr.21: 36223336-36223336 on Build GRCh38
6270488 c.1281G>T p.E427D Chr.21: 36223261-36223261 on Build GRCh38
1632472 c.1349G>A p.R450K Chr.21: 36223329-36223329 on Build GRCh38
137735 c.1294G>A p.V432I Chr.21: 36223274-36223274 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753456671 Chr.21: 36223273-36223273 on Build GRCh38
rs755543700 Chr.21: 36223185-36223185 on Build GRCh38
rs767331304 Chr.21: 36223181-36223181 on Build GRCh38
rs999163390 Chr.21: 36223267-36223267 on Build GRCh38
rs768834173 Chr.21: 36223206-36223206 on Build GRCh38
rs79458173 Chr.21: 36223172-36223172 on Build GRCh38
rs931470983 Chr.21: 36223173-36223174 on Build GRCh38
rs1022979835 Chr.21: 36223332-36223332 on Build GRCh38
rs182957792 Chr.21: 36223340-36223340 on Build GRCh38
rs539742056 Chr.21: 36223195-36223195 on Build GRCh38
rs752246408 Chr.21: 36223265-36223265 on Build GRCh38
rs866075140 Chr.21: 36223248-36223248 on Build GRCh38
rs769700465 Chr.21: 36223213-36223213 on Build GRCh38
rs201323448 Chr.21: 36223371-36223371 on Build GRCh38
rs752091694 Chr.21: 36223309-36223309 on Build GRCh38
rs557375780 Chr.21: 36223170-36223170 on Build GRCh38
rs748248615 Chr.21: 36223342-36223342 on Build GRCh38
rs780577213 Chr.21: 36223301-36223301 on Build GRCh38
rs767455893 Chr.21: 36223263-36223263 on Build GRCh38
rs199867717 Chr.21: 36223264-36223264 on Build GRCh38
rs542364079 Chr.21: 36223347-36223347 on Build GRCh38
rs371233048 Chr.21: 36223310-36223310 on Build GRCh38
rs1030295728 Chr.21: 36223212-36223212 on Build GRCh38
rs972795744 Chr.21: 36223171-36223171 on Build GRCh38
rs759448039 Chr.21: 36223257-36223257 on Build GRCh38
rs774268682 Chr.21: 36223253-36223253 on Build GRCh38
rs758352263 Chr.21: 36223189-36223189 on Build GRCh38
rs749436726 Chr.21: 36223358-36223358 on Build GRCh38
rs777365518 Chr.21: 36223192-36223192 on Build GRCh38
rs770964088 Chr.21: 36223365-36223365 on Build GRCh38
rs764951278 Chr.21: 36223272-36223272 on Build GRCh38
rs1004769835 Chr.21: 36223234-36223234 on Build GRCh38
rs766480016 Chr.21: 36223224-36223224 on Build GRCh38
rs747937869 Chr.21: 36223202-36223203 on Build GRCh38
rs373877576 Chr.21: 36223357-36223357 on Build GRCh38
rs773342018 Chr.21: 36223217-36223217 on Build GRCh38
rs374259127 Chr.21: 36223207-36223207 on Build GRCh38
rs1012969368 Chr.21: 36223322-36223322 on Build GRCh38
rs780394999 Chr.21: 36223196-36223196 on Build GRCh38
rs747421621 Chr.21: 36223202-36223202 on Build GRCh38
rs372238128 Chr.21: 36223189-36223189 on Build GRCh38
rs771677804 Chr.21: 36223257-36223258 on Build GRCh38
rs774922629 Chr.21: 36223274-36223274 on Build GRCh38
rs1017066101 Chr.21: 36223254-36223254 on Build GRCh38
rs373096935 Chr.21: 36223182-36223182 on Build GRCh38
rs112231366 Chr.21: 36223223-36223223 on Build GRCh38

View Full Product Details