Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 12825857-12826094 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX47
Entrez Gene ID
51202
Gene Name
DEAD-box helicase 47
Gene Aliases
E4-DBP, HQ0256, MSTP162, RRP3
UniGene
Hs.719938
Gene Chromosome Location
Chr.12: 12813346-12829981 on Build GRCh38
Transcripts
NM_016355.3 NM_201224.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2228082 c.1069G>C p.A357P Chr.12: 12826033-12826033 on Build GRCh38
1360050 c.1080C>T p.S360S Chr.12: 12826044-12826044 on Build GRCh38
5951368 c.1058G>A p.R353Q Chr.12: 12826022-12826022 on Build GRCh38
5620651 c.1088C>T p.A363V Chr.12: 12826052-12826052 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1003109773 Chr.12: 12825944-12825944 on Build GRCh38
rs764086548 Chr.12: 12825985-12825985 on Build GRCh38
rs150917198 Chr.12: 12826026-12826026 on Build GRCh38
rs769181617 Chr.12: 12826034-12826034 on Build GRCh38
rs769510425 Chr.12: 12826049-12826049 on Build GRCh38
rs147570642 Chr.12: 12825897-12825897 on Build GRCh38
rs1037594016 Chr.12: 12826005-12826005 on Build GRCh38
rs192161776 Chr.12: 12825967-12825967 on Build GRCh38
rs751687016 Chr.12: 12825988-12825988 on Build GRCh38
rs749585540 Chr.12: 12826030-12826030 on Build GRCh38
rs979865469 Chr.12: 12825943-12825944 on Build GRCh38
rs912376072 Chr.12: 12826004-12826004 on Build GRCh38
rs781518128 Chr.12: 12825998-12825998 on Build GRCh38
rs775459025 Chr.12: 12826054-12826054 on Build GRCh38
rs778765032 Chr.12: 12826016-12826016 on Build GRCh38
rs1027520290 Chr.12: 12825971-12825971 on Build GRCh38
rs753593371 Chr.12: 12825902-12825902 on Build GRCh38
rs762790654 Chr.12: 12826056-12826056 on Build GRCh38
rs758417123 Chr.12: 12825978-12825978 on Build GRCh38
rs370292279 Chr.12: 12825974-12825974 on Build GRCh38
rs746577860 Chr.12: 12826038-12826038 on Build GRCh38
rs923616212 Chr.12: 12826040-12826040 on Build GRCh38
rs936295606 Chr.12: 12826059-12826059 on Build GRCh38
rs1023132025 Chr.12: 12825934-12825934 on Build GRCh38
rs760062331 Chr.12: 12825952-12825952 on Build GRCh38
rs764144574 Chr.12: 12826063-12826063 on Build GRCh38
rs759440076 Chr.12: 12825963-12825963 on Build GRCh38
rs367616019 Chr.12: 12825959-12825959 on Build GRCh38
rs79487375 Chr.12: 12825903-12825903 on Build GRCh38
rs757266709 Chr.12: 12825991-12825991 on Build GRCh38
rs781695222 Chr.12: 12825882-12825882 on Build GRCh38
rs772782271 Chr.12: 12825951-12825951 on Build GRCh38
rs746073969 Chr.12: 12826012-12826012 on Build GRCh38
rs756530038 Chr.12: 12826021-12826021 on Build GRCh38
rs770311332 Chr.12: 12826039-12826039 on Build GRCh38
rs776407507 Chr.12: 12826043-12826043 on Build GRCh38
rs200552579 Chr.12: 12826044-12826044 on Build GRCh38
rs188907506 Chr.12: 12825941-12825941 on Build GRCh38

View Full Product Details