Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 87059073-87059346 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLUD1
Entrez Gene ID
2746
Gene Name
glutamate dehydrogenase 1
Gene Aliases
GDH, GDH1, GLUD
UniGene
Hs.500409
Gene Chromosome Location
Chr.10: 87050202-87095047 on Build GRCh38
Transcripts
NM_001318900.1 NM_001318901.1 NM_001318902.1 NM_001318904.1 NM_001318905.1 NM_001318906.1 NM_005271.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3807878 c.1385C>A p.S462Y Chr.10: 87059167-87059167 on Build GRCh38
5485399 c.1358G>A p.R453H Chr.10: 87059194-87059194 on Build GRCh38
4954412 c.1386T>C p.S462S Chr.10: 87059166-87059166 on Build GRCh38
7348085 c.1336C>A p.L446I Chr.10: 87059216-87059216 on Build GRCh38
4978568 c.1342C>T p.H448Y Chr.10: 87059210-87059210 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772424574 Chr.10: 87059101-87059101 on Build GRCh38
rs375751878 Chr.10: 87059318-87059318 on Build GRCh38
rs202067232 Chr.10: 87059140-87059140 on Build GRCh38
rs142635640 Chr.10: 87059127-87059127 on Build GRCh38
rs146034815 Chr.10: 87059128-87059128 on Build GRCh38
rs938285596 Chr.10: 87059310-87059310 on Build GRCh38
rs765785913 Chr.10: 87059303-87059308 on Build GRCh38
rs868589833 Chr.10: 87059273-87059273 on Build GRCh38
rs957243299 Chr.10: 87059216-87059216 on Build GRCh38
rs746026416 Chr.10: 87059100-87059100 on Build GRCh38
rs772331471 Chr.10: 87059305-87059305 on Build GRCh38
rs371156720 Chr.10: 87059142-87059142 on Build GRCh38
rs201376212 Chr.10: 87059277-87059277 on Build GRCh38
rs1031986279 Chr.10: 87059202-87059202 on Build GRCh38
rs577763265 Chr.10: 87059112-87059112 on Build GRCh38
rs754345029 Chr.10: 87059182-87059182 on Build GRCh38
rs756106006 Chr.10: 87059291-87059292 on Build GRCh38
rs757850216 Chr.10: 87059244-87059244 on Build GRCh38
rs374548261 Chr.10: 87059279-87059279 on Build GRCh38
rs1018744872 Chr.10: 87059104-87059104 on Build GRCh38
rs768540352 Chr.10: 87059123-87059123 on Build GRCh38
rs895203336 Chr.10: 87059114-87059114 on Build GRCh38
rs756400866 Chr.10: 87059178-87059178 on Build GRCh38
rs751273734 Chr.10: 87059148-87059148 on Build GRCh38
rs780112936 Chr.10: 87059308-87059308 on Build GRCh38
rs778228652 Chr.10: 87059181-87059181 on Build GRCh38
rs200282708 Chr.10: 87059232-87059232 on Build GRCh38
rs760369226 Chr.10: 87059118-87059118 on Build GRCh38
rs971143389 Chr.10: 87059242-87059242 on Build GRCh38

View Full Product Details