Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 121090964-121091219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBC1D32
Entrez Gene ID
221322
Gene Name
TBC1 domain family member 32
Gene Aliases
BROMI, C6orf170, C6orf171, bA301B7.2, bA57L9.1, dJ310J6.1
UniGene
Hs.121396
Gene Chromosome Location
Chr.6: 121079494-121335382 on Build GRCh38
Transcripts
NM_152730.5 NR_104452.1 XM_005266861.2 XM_011535569.1 XM_011535570.1 XM_011535571.2 XM_011535574.1 XM_011535580.2 XM_017010397.1 XM_017010398.1 XM_017010399.1 XM_017010400.1 XM_017010401.1 XM_017010404.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5589937 c.3497G>A p.W1166* Chr.6: 121091010-121091010 on Build GRCh38
6375801 c.3475C>G p.Q1159E Chr.6: 121091032-121091032 on Build GRCh38
4961919 c.3466-4A>T p.? Chr.6: 121091045-121091045 on Build GRCh38
4773650 c.3466-5delT p.? Chr.6: 121091046-121091046 on Build GRCh38
3857937 c.3485C>T p.T1162I Chr.6: 121091022-121091022 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759733610 Chr.6: 121091052-121091052 on Build GRCh38
rs776487629 Chr.6: 121091026-121091026 on Build GRCh38
rs1030018520 Chr.6: 121091163-121091163 on Build GRCh38
rs751211165 Chr.6: 121091020-121091020 on Build GRCh38
rs888646123 Chr.6: 121091042-121091043 on Build GRCh38
rs1017388369 Chr.6: 121091062-121091062 on Build GRCh38
rs749173310 Chr.6: 121091050-121091051 on Build GRCh38
rs994830643 Chr.6: 121090995-121090995 on Build GRCh38
rs549791728 Chr.6: 121091045-121091046 on Build GRCh38
rs144653188 Chr.6: 121091060-121091060 on Build GRCh38
rs750106845 Chr.6: 121091091-121091091 on Build GRCh38
rs761280562 Chr.6: 121091067-121091067 on Build GRCh38
rs964359789 Chr.6: 121091096-121091096 on Build GRCh38
rs1023650160 Chr.6: 121091125-121091125 on Build GRCh38
rs558860359 Chr.6: 121091080-121091080 on Build GRCh38
rs746970154 Chr.6: 121090997-121090997 on Build GRCh38
rs772313204 Chr.6: 121091058-121091058 on Build GRCh38
rs768418401 Chr.6: 121091021-121091021 on Build GRCh38
rs779996106 Chr.6: 121091048-121091060 on Build GRCh38
rs772321070 Chr.6: 121091094-121091094 on Build GRCh38
rs397887772 Chr.6: 121091056-121091057 on Build GRCh38
rs751888049 Chr.6: 121091146-121091146 on Build GRCh38
rs74884690 Chr.6: 121091045-121091045 on Build GRCh38
rs747892622 Chr.6: 121091030-121091030 on Build GRCh38
rs778644325 Chr.6: 121091058-121091059 on Build GRCh38
rs764502399 Chr.6: 121091069-121091069 on Build GRCh38
rs755511050 Chr.6: 121091046-121091048 on Build GRCh38
rs190741035 Chr.6: 121091071-121091071 on Build GRCh38
rs6931402 Chr.6: 121091147-121091147 on Build GRCh38
rs774693730 Chr.6: 121091046-121091046 on Build GRCh38
rs756930507 Chr.6: 121091046-121091046 on Build GRCh38
rs35529405 Chr.6: 121091100-121091100 on Build GRCh38
rs748383430 Chr.6: 121091060-121091061 on Build GRCh38
rs974204033 Chr.6: 121091171-121091171 on Build GRCh38
rs764977249 Chr.6: 121091083-121091083 on Build GRCh38
rs775665467 Chr.6: 121091063-121091063 on Build GRCh38
rs771243654 Chr.6: 121091036-121091036 on Build GRCh38
rs768484461 Chr.6: 121091056-121091058 on Build GRCh38

View Full Product Details