Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64947165-64947432 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAJIN
Entrez Gene ID
283129
Gene Name
membrane anchored junction protein
Gene Aliases
C11orf85
UniGene
Hs.567793
Gene Chromosome Location
Chr.11: 64938230-64972104 on Build GRCh38
Transcripts
NM_001037225.2 NM_001300803.1 NM_001318808.1 XM_005273918.2 XM_006718502.2 XM_006718503.3 XM_006718504.2 XM_006718505.2 XM_011544940.2 XM_011544941.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4035302 c.416T>C p.M139T Chr.11: 64947431-64947431 on Build GRCh38
6310839 c.442C>A p.P148T Chr.11: 64947405-64947405 on Build GRCh38
4891149 c.99G>A p.M33I Chr.11: 64947430-64947430 on Build GRCh38
4891148 c.417G>A p.M139I Chr.11: 64947430-64947430 on Build GRCh38
5782719 c.120C>T p.D40D Chr.11: 64947409-64947409 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764323891 Chr.11: 64947336-64947336 on Build GRCh38
rs752690898 Chr.11: 64947376-64947376 on Build GRCh38
rs527934328 Chr.11: 64947295-64947295 on Build GRCh38
rs7482817 Chr.11: 64947308-64947308 on Build GRCh38
rs765236733 Chr.11: 64947364-64947364 on Build GRCh38
rs149450150 Chr.11: 64947396-64947396 on Build GRCh38
rs201309929 Chr.11: 64947327-64947327 on Build GRCh38
rs1006424349 Chr.11: 64947289-64947289 on Build GRCh38
rs756956874 Chr.11: 64947397-64947397 on Build GRCh38
rs780767889 Chr.11: 64947389-64947389 on Build GRCh38
rs753702004 Chr.11: 64947349-64947349 on Build GRCh38
rs956655937 Chr.11: 64947380-64947380 on Build GRCh38
rs138095675 Chr.11: 64947323-64947323 on Build GRCh38
rs4320955 Chr.11: 64947212-64947212 on Build GRCh38
rs568480613 Chr.11: 64947311-64947311 on Build GRCh38
rs551356904 Chr.11: 64947234-64947234 on Build GRCh38
rs1046731346 Chr.11: 64947366-64947366 on Build GRCh38
rs1030340157 Chr.11: 64947316-64947316 on Build GRCh38
rs370936071 Chr.11: 64947341-64947341 on Build GRCh38
rs757900939 Chr.11: 64947382-64947382 on Build GRCh38
rs898099212 Chr.11: 64947310-64947310 on Build GRCh38
rs754808374 Chr.11: 64947358-64947358 on Build GRCh38
rs560266308 Chr.11: 64947340-64947340 on Build GRCh38
rs145232114 Chr.11: 64947409-64947409 on Build GRCh38
rs538298585 Chr.11: 64947348-64947348 on Build GRCh38
rs531635963 Chr.11: 64947233-64947233 on Build GRCh38
rs781370247 Chr.11: 64947406-64947406 on Build GRCh38
rs140240334 Chr.11: 64947408-64947408 on Build GRCh38
rs762912885 Chr.11: 64947257-64947257 on Build GRCh38
rs531786156 Chr.11: 64947241-64947241 on Build GRCh38
rs145543501 Chr.11: 64947199-64947199 on Build GRCh38
rs763136147 Chr.11: 64947335-64947335 on Build GRCh38
rs373891467 Chr.11: 64947321-64947321 on Build GRCh38

View Full Product Details