Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 35863311-35863584 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HEATR9
Entrez Gene ID
256957
Gene Name
HEAT repeat containing 9
Gene Aliases
C17orf66
UniGene
Hs.514820
Gene Chromosome Location
Chr.17: 35854946-35868891 on Build GRCh38
Transcripts
NM_001321395.1 NM_152781.3 NR_135630.1 XM_005278233.2 XM_005278235.1 XM_006721802.3 XM_011524586.2 XM_017024424.1

SNP Detail

SNP ID Chromosome Location
rs775258843 Chr.17: 35863462-35863462 on Build GRCh38
rs183997741 Chr.17: 35863520-35863520 on Build GRCh38
rs757040296 Chr.17: 35863556-35863556 on Build GRCh38
rs762535276 Chr.17: 35863464-35863464 on Build GRCh38
rs147813167 Chr.17: 35863558-35863558 on Build GRCh38
rs115881734 Chr.17: 35863512-35863512 on Build GRCh38
rs746737497 Chr.17: 35863454-35863454 on Build GRCh38
rs770225186 Chr.17: 35863449-35863449 on Build GRCh38
rs768315774 Chr.17: 35863481-35863481 on Build GRCh38
rs532626226 Chr.17: 35863418-35863418 on Build GRCh38
rs62078235 Chr.17: 35863527-35863527 on Build GRCh38
rs377475394 Chr.17: 35863530-35863530 on Build GRCh38
rs765382917 Chr.17: 35863529-35863529 on Build GRCh38
rs978101702 Chr.17: 35863379-35863379 on Build GRCh38
rs774648611 Chr.17: 35863496-35863499 on Build GRCh38
rs975608961 Chr.17: 35863354-35863354 on Build GRCh38
rs753224534 Chr.17: 35863446-35863446 on Build GRCh38
rs929218591 Chr.17: 35863521-35863521 on Build GRCh38
rs771362515 Chr.17: 35863525-35863525 on Build GRCh38
rs953157779 Chr.17: 35863365-35863365 on Build GRCh38
rs552237376 Chr.17: 35863513-35863513 on Build GRCh38
rs773855758 Chr.17: 35863515-35863515 on Build GRCh38
rs781092264 Chr.17: 35863457-35863457 on Build GRCh38
rs552396184 Chr.17: 35863455-35863455 on Build GRCh38
rs769154410 Chr.17: 35863460-35863460 on Build GRCh38
rs767733905 Chr.17: 35863445-35863445 on Build GRCh38
rs755157020 Chr.17: 35863522-35863522 on Build GRCh38
rs191852266 Chr.17: 35863360-35863360 on Build GRCh38
rs920303541 Chr.17: 35863358-35863358 on Build GRCh38
rs760853295 Chr.17: 35863517-35863517 on Build GRCh38
rs986342902 Chr.17: 35863388-35863388 on Build GRCh38
rs560407403 Chr.17: 35863356-35863356 on Build GRCh38
rs774861589 Chr.17: 35863484-35863484 on Build GRCh38
rs570357362 Chr.17: 35863374-35863374 on Build GRCh38
rs756734621 Chr.17: 35863470-35863470 on Build GRCh38
rs745449063 Chr.17: 35863458-35863458 on Build GRCh38

View Full Product Details