Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 111649329-111649590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC25
Entrez Gene ID
548645
Gene Name
DnaJ heat shock protein family (Hsp40) member C25
Gene Aliases
bA16L21.2.1
UniGene
Hs.534196
Gene Chromosome Location
Chr.9: 111631352-111654351 on Build GRCh38
Transcripts
NM_001015882.2 NR_037148.1 NM_004125.3

Gene Symbol
DNAJC25-GNG10
Entrez Gene ID
552891
Gene Name
DNAJC25-GNG10 readthrough
Gene Aliases
-
UniGene
Hs.534196
Gene Chromosome Location
Chr.9: 111631352-111670246 on Build GRCh38
Transcripts
NM_001015882.2 NR_037148.1 NM_004125.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1204210 c.607G>T p.E203* Chr.9: 111649570-111649570 on Build GRCh38
4163214 c.616A>T p.K206* Chr.9: 111649579-111649579 on Build GRCh38
3169975 c.614G>A p.G205D Chr.9: 111649577-111649577 on Build GRCh38
4163213 c.615C>A p.G205G Chr.9: 111649578-111649578 on Build GRCh38
1104076 c.559_560insA p.A188fs*29 Chr.9: 111649522-111649523 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs552447394 Chr.9: 111649450-111649450 on Build GRCh38
rs368887646 Chr.9: 111649489-111649489 on Build GRCh38
rs983684481 Chr.9: 111649424-111649424 on Build GRCh38
rs374845005 Chr.9: 111649478-111649478 on Build GRCh38
rs537603658 Chr.9: 111649501-111649501 on Build GRCh38
rs776791144 Chr.9: 111649503-111649503 on Build GRCh38
rs760951719 Chr.9: 111649500-111649500 on Build GRCh38
rs921977631 Chr.9: 111649541-111649541 on Build GRCh38
rs538717472 Chr.9: 111649429-111649429 on Build GRCh38
rs754682829 Chr.9: 111649525-111649525 on Build GRCh38
rs780670002 Chr.9: 111649551-111649551 on Build GRCh38
rs372256683 Chr.9: 111649517-111649517 on Build GRCh38
rs1002922095 Chr.9: 111649442-111649442 on Build GRCh38
rs766771850 Chr.9: 111649518-111649518 on Build GRCh38
rs193192759 Chr.9: 111649389-111649389 on Build GRCh38
rs763551475 Chr.9: 111649526-111649526 on Build GRCh38
rs758802430 Chr.9: 111649422-111649422 on Build GRCh38
rs750942878 Chr.9: 111649516-111649516 on Build GRCh38
rs375241958 Chr.9: 111649430-111649430 on Build GRCh38
rs746669881 Chr.9: 111649432-111649432 on Build GRCh38
rs754520350 Chr.9: 111649438-111649438 on Build GRCh38
rs147227293 Chr.9: 111649365-111649365 on Build GRCh38
rs773225801 Chr.9: 111649426-111649426 on Build GRCh38
rs769756835 Chr.9: 111649454-111649454 on Build GRCh38
rs554851029 Chr.9: 111649508-111649508 on Build GRCh38
rs752247206 Chr.9: 111649559-111649559 on Build GRCh38
rs760750830 Chr.9: 111649530-111649531 on Build GRCh38
rs751812581 Chr.9: 111649519-111649519 on Build GRCh38
rs1032205178 Chr.9: 111649476-111649476 on Build GRCh38
rs535422079 Chr.9: 111649443-111649443 on Build GRCh38
rs368856452 Chr.9: 111649377-111649377 on Build GRCh38
rs371506049 Chr.9: 111649477-111649477 on Build GRCh38
rs367873033 Chr.9: 111649468-111649468 on Build GRCh38

View Full Product Details