Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 201274792-201275051 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP8
Entrez Gene ID
841
Gene Name
caspase 8
Gene Aliases
ALPS2B, CAP4, Casp-8, FLICE, MACH, MCH5
UniGene
Hs.599762
Gene Chromosome Location
Chr.2: 201233443-201287711 on Build GRCh38
Transcripts
NM_001080124.1 NM_001080125.1 NM_001228.4 NM_033355.3 NM_033356.3 NM_033358.3 NR_111983.1 XM_005246885.1 XM_005246886.1 XM_005246887.1 XM_005246888.1 XM_005246889.1 XM_005246890.3 XM_005246891.4 XM_005246892.1 XM_005246893.2 XM_005246894.3 XM_005246895.2 XM_006712789.1 XM_006712790.3 XM_006712793.2 XM_011511969.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5007626 c.671C>T p.T224I Chr.2: 201274913-201274913 on Build GRCh38
3721143 c.698A>G p.D233G Chr.2: 201274940-201274940 on Build GRCh38
3798499 c.809C>G p.S270C Chr.2: 201274925-201274925 on Build GRCh38
6926966 c.649G>T p.E217* Chr.2: 201274891-201274891 on Build GRCh38
4450515 c.647-1delG p.? Chr.2: 201274888-201274888 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756817274 Chr.2: 201274877-201274877 on Build GRCh38
rs766429077 Chr.2: 201274960-201274960 on Build GRCh38
rs971778876 Chr.2: 201274923-201274923 on Build GRCh38
rs759966804 Chr.2: 201274986-201274986 on Build GRCh38
rs918397149 Chr.2: 201274822-201274822 on Build GRCh38
rs374619190 Chr.2: 201274825-201274825 on Build GRCh38
rs894549732 Chr.2: 201274866-201274866 on Build GRCh38
rs761692687 Chr.2: 201274938-201274938 on Build GRCh38
rs779769497 Chr.2: 201274908-201274908 on Build GRCh38
rs771815245 Chr.2: 201274942-201274942 on Build GRCh38
rs764103691 Chr.2: 201274975-201274976 on Build GRCh38
rs372597885 Chr.2: 201274994-201274994 on Build GRCh38
rs535411498 Chr.2: 201274975-201274975 on Build GRCh38
rs765761798 Chr.2: 201274990-201274990 on Build GRCh38
rs749179894 Chr.2: 201274920-201274920 on Build GRCh38
rs777498788 Chr.2: 201274976-201274976 on Build GRCh38
rs35976359 Chr.2: 201274948-201274948 on Build GRCh38
rs200861691 Chr.2: 201274996-201274996 on Build GRCh38
rs146816437 Chr.2: 201274933-201274933 on Build GRCh38
rs577264458 Chr.2: 201274903-201274903 on Build GRCh38
rs368079846 Chr.2: 201274989-201274989 on Build GRCh38
rs553817414 Chr.2: 201274992-201274992 on Build GRCh38
rs555978298 Chr.2: 201274874-201274874 on Build GRCh38
rs755533786 Chr.2: 201274853-201274853 on Build GRCh38
rs35508730 Chr.2: 201275016-201275017 on Build GRCh38
rs989271581 Chr.2: 201274997-201274997 on Build GRCh38
rs563444320 Chr.2: 201274943-201274943 on Build GRCh38
rs769315372 Chr.2: 201274899-201274899 on Build GRCh38
rs953248061 Chr.2: 201275011-201275011 on Build GRCh38
rs369771634 Chr.2: 201274847-201274847 on Build GRCh38
rs377152347 Chr.2: 201274878-201274878 on Build GRCh38
rs553241803 Chr.2: 201274896-201274896 on Build GRCh38
rs779188502 Chr.2: 201274868-201274868 on Build GRCh38
rs768721175 Chr.2: 201274928-201274928 on Build GRCh38
rs370400442 Chr.2: 201274959-201274959 on Build GRCh38
rs879187393 Chr.2: 201274836-201274836 on Build GRCh38
rs181699083 Chr.2: 201274851-201274851 on Build GRCh38
rs915093667 Chr.2: 201274999-201274999 on Build GRCh38
rs62191536 Chr.2: 201274972-201274972 on Build GRCh38
rs753925201 Chr.2: 201274966-201274966 on Build GRCh38

View Full Product Details