Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 30730639-30730898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN13
Entrez Gene ID
92291
Gene Name
calpain 13
Gene Aliases
-
UniGene
Hs.660911
Gene Chromosome Location
Chr.2: 30610382-30809527 on Build GRCh38
Transcripts
NM_144575.2 XM_011533159.2 XM_011533160.2 XM_011533161.2 XM_017005264.1 XM_017005265.1 XM_017005266.1

SNP Detail

SNP ID Chromosome Location
rs909895653 Chr.2: 30730855-30730855 on Build GRCh38
rs895389473 Chr.2: 30730664-30730664 on Build GRCh38
rs999688879 Chr.2: 30730692-30730692 on Build GRCh38
rs746103813 Chr.2: 30730856-30730856 on Build GRCh38
rs748932423 Chr.2: 30730722-30730722 on Build GRCh38
rs1021964296 Chr.2: 30730678-30730678 on Build GRCh38
rs770460303 Chr.2: 30730740-30730740 on Build GRCh38
rs531637395 Chr.2: 30730813-30730813 on Build GRCh38
rs766487024 Chr.2: 30730824-30730824 on Build GRCh38
rs377762647 Chr.2: 30730821-30730821 on Build GRCh38
rs148437016 Chr.2: 30730719-30730719 on Build GRCh38
rs894999519 Chr.2: 30730738-30730738 on Build GRCh38
rs1042117908 Chr.2: 30730694-30730694 on Build GRCh38
rs764769112 Chr.2: 30730777-30730777 on Build GRCh38
rs761994026 Chr.2: 30730793-30730793 on Build GRCh38
rs1030709727 Chr.2: 30730696-30730696 on Build GRCh38
rs761110822 Chr.2: 30730771-30730771 on Build GRCh38
rs775975059 Chr.2: 30730746-30730746 on Build GRCh38
rs771838885 Chr.2: 30730722-30730722 on Build GRCh38
rs985460248 Chr.2: 30730787-30730787 on Build GRCh38
rs538002071 Chr.2: 30730708-30730708 on Build GRCh38
rs903633312 Chr.2: 30730685-30730685 on Build GRCh38
rs962703871 Chr.2: 30730871-30730871 on Build GRCh38
rs539587295 Chr.2: 30730669-30730669 on Build GRCh38
rs765308384 Chr.2: 30730805-30730805 on Build GRCh38
rs1055329105 Chr.2: 30730721-30730721 on Build GRCh38
rs950691780 Chr.2: 30730861-30730861 on Build GRCh38
rs953294094 Chr.2: 30730778-30730778 on Build GRCh38
rs1011944727 Chr.2: 30730667-30730667 on Build GRCh38
rs751412346 Chr.2: 30730834-30730834 on Build GRCh38
rs1028894553 Chr.2: 30730764-30730764 on Build GRCh38
rs750639017 Chr.2: 30730812-30730812 on Build GRCh38
rs553331143 Chr.2: 30730679-30730679 on Build GRCh38
rs566777825 Chr.2: 30730697-30730697 on Build GRCh38

View Full Product Details