Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10929282-10929547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf52
Entrez Gene ID
90580
Gene Name
chromosome 19 open reading frame 52
Gene Aliases
-
UniGene
Hs.164026
Gene Chromosome Location
Chr.19: 10928748-10930241 on Build GRCh38
Transcripts
NM_138358.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4154036 c.574C>T p.L192L Chr.19: 10929493-10929493 on Build GRCh38
6418980 c.373G>A p.V125I Chr.19: 10929292-10929292 on Build GRCh38
4580599 c.463C>A p.R155S Chr.19: 10929382-10929382 on Build GRCh38
6576281 c.525G>A p.L175L Chr.19: 10929444-10929444 on Build GRCh38
3796585 c.556G>T p.D186Y Chr.19: 10929475-10929475 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762059274 Chr.19: 10929502-10929502 on Build GRCh38
rs747744588 Chr.19: 10929468-10929468 on Build GRCh38
rs1005301767 Chr.19: 10929504-10929504 on Build GRCh38
rs1016784363 Chr.19: 10929356-10929356 on Build GRCh38
rs764222201 Chr.19: 10929491-10929491 on Build GRCh38
rs1027960252 Chr.19: 10929495-10929495 on Build GRCh38
rs148467140 Chr.19: 10929490-10929490 on Build GRCh38
rs368933242 Chr.19: 10929393-10929393 on Build GRCh38
rs770577761 Chr.19: 10929422-10929430 on Build GRCh38
rs373453656 Chr.19: 10929408-10929408 on Build GRCh38
rs751506909 Chr.19: 10929510-10929510 on Build GRCh38
rs779634694 Chr.19: 10929450-10929450 on Build GRCh38
rs763344718 Chr.19: 10929383-10929383 on Build GRCh38
rs750389431 Chr.19: 10929438-10929438 on Build GRCh38
rs148628785 Chr.19: 10929487-10929487 on Build GRCh38
rs750193226 Chr.19: 10929508-10929508 on Build GRCh38
rs200318904 Chr.19: 10929329-10929329 on Build GRCh38
rs539014018 Chr.19: 10929423-10929423 on Build GRCh38
rs768140430 Chr.19: 10929381-10929381 on Build GRCh38
rs538123098 Chr.19: 10929384-10929384 on Build GRCh38
rs755111488 Chr.19: 10929318-10929318 on Build GRCh38
rs777646969 Chr.19: 10929336-10929336 on Build GRCh38
rs752972395 Chr.19: 10929327-10929327 on Build GRCh38
rs767440748 Chr.19: 10929321-10929321 on Build GRCh38
rs555565275 Chr.19: 10929516-10929516 on Build GRCh38
rs201384798 Chr.19: 10929362-10929362 on Build GRCh38
rs776213771 Chr.19: 10929471-10929473 on Build GRCh38
rs760114809 Chr.19: 10929486-10929486 on Build GRCh38
rs757088126 Chr.19: 10929342-10929342 on Build GRCh38
rs756197771 Chr.19: 10929417-10929417 on Build GRCh38
rs754501820 Chr.19: 10929514-10929514 on Build GRCh38
rs757173956 Chr.19: 10929430-10929430 on Build GRCh38
rs752026693 Chr.19: 10929311-10929311 on Build GRCh38
rs961462422 Chr.19: 10929380-10929380 on Build GRCh38
rs776034474 Chr.19: 10929489-10929489 on Build GRCh38
rs774795534 Chr.19: 10929367-10929367 on Build GRCh38
rs778791266 Chr.19: 10929434-10929434 on Build GRCh38
rs765153717 Chr.19: 10929505-10929505 on Build GRCh38
rs147145708 Chr.19: 10929403-10929403 on Build GRCh38
rs750912802 Chr.19: 10929509-10929509 on Build GRCh38
rs927398022 Chr.19: 10929404-10929404 on Build GRCh38
rs745785163 Chr.19: 10929359-10929359 on Build GRCh38
rs202159235 Chr.19: 10929501-10929501 on Build GRCh38
rs761237926 Chr.19: 10929526-10929526 on Build GRCh38
rs549144776 Chr.19: 10929308-10929308 on Build GRCh38
rs746706261 Chr.19: 10929372-10929372 on Build GRCh38
rs764265303 Chr.19: 10929420-10929420 on Build GRCh38
rs34083039 Chr.19: 10929451-10929451 on Build GRCh38
rs748615198 Chr.19: 10929334-10929346 on Build GRCh38
rs766918046 Chr.19: 10929309-10929309 on Build GRCh38
rs779062829 Chr.19: 10929351-10929351 on Build GRCh38
rs950069739 Chr.19: 10929401-10929401 on Build GRCh38
rs760807134 Chr.19: 10929519-10929519 on Build GRCh38
rs747832687 Chr.19: 10929520-10929520 on Build GRCh38
rs138904551 Chr.19: 10929446-10929446 on Build GRCh38
rs763468858 Chr.19: 10929304-10929304 on Build GRCh38
rs771293577 Chr.19: 10929475-10929475 on Build GRCh38
rs540484501 Chr.19: 10929382-10929382 on Build GRCh38
rs752675227 Chr.19: 10929411-10929411 on Build GRCh38
rs766723667 Chr.19: 10929400-10929400 on Build GRCh38
rs780887647 Chr.19: 10929460-10929460 on Build GRCh38
rs550170903 Chr.19: 10929385-10929385 on Build GRCh38
rs749145423 Chr.19: 10929338-10929338 on Build GRCh38
rs777386752 Chr.19: 10929521-10929521 on Build GRCh38
rs868567284 Chr.19: 10929457-10929457 on Build GRCh38
rs376399324 Chr.19: 10929478-10929478 on Build GRCh38
rs1028055153 Chr.19: 10929317-10929317 on Build GRCh38
rs917160991 Chr.19: 10929397-10929397 on Build GRCh38
rs371039420 Chr.19: 10929513-10929513 on Build GRCh38

View Full Product Details