Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218662630-218662883 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCS1L
Entrez Gene ID
617
Gene Name
BCS1 homolog, ubiquinol-cytochrome c reductase complex chaperone
Gene Aliases
BCS, BCS1, BJS, FLNMS, GRACILE, Hs.6719, MC3DN1, PTD, h-BCS, h-BCS1
UniGene
Hs.471401
Gene Chromosome Location
Chr.2: 218659656-218663443 on Build GRCh38
Transcripts
NM_001079866.1 NM_001257342.1 NM_001257343.1 NM_001257344.1 NM_001318836.1 NM_001320717.1 NM_004328.4 XM_005246748.2 XM_005246749.3 XM_006712678.1 XM_017004631.1 XM_017004632.1 XM_017004633.1 XM_017004634.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7226471 c.885G>A p.V295V Chr.2: 218662675-218662675 on Build GRCh38
1669757 c.871C>T p.R291* Chr.2: 218662661-218662661 on Build GRCh38
5551983 c.853G>C p.D285H Chr.2: 218662643-218662643 on Build GRCh38
6575680 c.865C>T p.L289F Chr.2: 218662655-218662655 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs766681582 Chr.2: 218662667-218662667 on Build GRCh38
rs754039539 Chr.2: 218662675-218662675 on Build GRCh38
rs890961719 Chr.2: 218662721-218662721 on Build GRCh38
rs773210885 Chr.2: 218662688-218662688 on Build GRCh38
rs779048403 Chr.2: 218662691-218662691 on Build GRCh38
rs565819523 Chr.2: 218662718-218662718 on Build GRCh38
rs887329900 Chr.2: 218662829-218662829 on Build GRCh38
rs114720751 Chr.2: 218662780-218662780 on Build GRCh38
rs1041524366 Chr.2: 218662719-218662719 on Build GRCh38
rs748288923 Chr.2: 218662695-218662695 on Build GRCh38
rs920238469 Chr.2: 218662707-218662707 on Build GRCh38
rs778635316 Chr.2: 218662715-218662715 on Build GRCh38
rs1057516346 Chr.2: 218662680-218662680 on Build GRCh38
rs763087998 Chr.2: 218662725-218662726 on Build GRCh38
rs758490202 Chr.2: 218662713-218662713 on Build GRCh38
rs775200203 Chr.2: 218662722-218662722 on Build GRCh38
rs944097929 Chr.2: 218662733-218662733 on Build GRCh38
rs750976781 Chr.2: 218662664-218662664 on Build GRCh38
rs917427682 Chr.2: 218662794-218662794 on Build GRCh38
rs554510882 Chr.2: 218662845-218662845 on Build GRCh38
rs1040191469 Chr.2: 218662768-218662768 on Build GRCh38
rs34933485 Chr.2: 218662686-218662687 on Build GRCh38
rs199791659 Chr.2: 218662820-218662820 on Build GRCh38
rs756477155 Chr.2: 218662666-218662666 on Build GRCh38
rs1044684262 Chr.2: 218662808-218662808 on Build GRCh38
rs974634821 Chr.2: 218662696-218662696 on Build GRCh38
rs768258849 Chr.2: 218662838-218662838 on Build GRCh38
rs914472798 Chr.2: 218662708-218662708 on Build GRCh38
rs748936714 Chr.2: 218662724-218662724 on Build GRCh38
rs755197780 Chr.2: 218662690-218662690 on Build GRCh38
rs948907642 Chr.2: 218662805-218662805 on Build GRCh38
rs940199665 Chr.2: 218662748-218662748 on Build GRCh38
rs769584762 Chr.2: 218662720-218662720 on Build GRCh38
rs201454788 Chr.2: 218662661-218662661 on Build GRCh38

View Full Product Details