Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7311305-7311568 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF5A
Entrez Gene ID
1984
Gene Name
eukaryotic translation initiation factor 5A
Gene Aliases
EIF-5A, EIF5A1, eIF5AI
UniGene
Hs.534314
Gene Chromosome Location
Chr.17: 7306999-7312463 on Build GRCh38
Transcripts
NM_001143760.1 NM_001143761.1 NM_001143762.1 NM_001970.4 XM_005256509.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4069262 c.428G>A p.R143H Chr.17: 7311417-7311417 on Build GRCh38
271237 c.302T>G p.L101R Chr.17: 7311381-7311381 on Build GRCh38
7129129 c.370G>C p.E124Q Chr.17: 7311449-7311449 on Build GRCh38
4069260 c.366T>C p.I122I Chr.17: 7311355-7311355 on Build GRCh38
4069264 c.467A>C p.K156T Chr.17: 7311456-7311456 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766260612 Chr.17: 7311346-7311346 on Build GRCh38
rs201073018 Chr.17: 7311460-7311460 on Build GRCh38
rs770708499 Chr.17: 7311521-7311521 on Build GRCh38
rs769345872 Chr.17: 7311500-7311500 on Build GRCh38
rs748647159 Chr.17: 7311499-7311499 on Build GRCh38
rs11658072 Chr.17: 7311505-7311505 on Build GRCh38
rs753811920 Chr.17: 7311448-7311448 on Build GRCh38
rs780331793 Chr.17: 7311485-7311485 on Build GRCh38
rs747407301 Chr.17: 7311488-7311488 on Build GRCh38
rs978360433 Chr.17: 7311538-7311538 on Build GRCh38
rs367629450 Chr.17: 7311343-7311343 on Build GRCh38
rs570238943 Chr.17: 7311345-7311345 on Build GRCh38
rs764706416 Chr.17: 7311342-7311342 on Build GRCh38
rs780108399 Chr.17: 7311471-7311472 on Build GRCh38
rs190355794 Chr.17: 7311515-7311515 on Build GRCh38
rs761412582 Chr.17: 7311338-7311338 on Build GRCh38
rs754892735 Chr.17: 7311388-7311388 on Build GRCh38
rs891726130 Chr.17: 7311394-7311394 on Build GRCh38
rs769102309 Chr.17: 7311495-7311495 on Build GRCh38
rs201657126 Chr.17: 7311466-7311466 on Build GRCh38
rs776078900 Chr.17: 7311335-7311335 on Build GRCh38
rs868508244 Chr.17: 7311493-7311493 on Build GRCh38
rs752647399 Chr.17: 7311532-7311532 on Build GRCh38
rs551402102 Chr.17: 7311510-7311510 on Build GRCh38
rs774244068 Chr.17: 7311527-7311527 on Build GRCh38
rs751411515 Chr.17: 7311380-7311380 on Build GRCh38
rs374999461 Chr.17: 7311496-7311496 on Build GRCh38
rs201013827 Chr.17: 7311541-7311541 on Build GRCh38
rs149769568 Chr.17: 7311451-7311451 on Build GRCh38
rs765304365 Chr.17: 7311545-7311545 on Build GRCh38
rs747458912 Chr.17: 7311331-7311331 on Build GRCh38
rs778898421 Chr.17: 7311457-7311457 on Build GRCh38
rs769157076 Chr.17: 7311334-7311334 on Build GRCh38
rs371720032 Chr.17: 7311409-7311409 on Build GRCh38
rs182952654 Chr.17: 7311528-7311528 on Build GRCh38
rs758634409 Chr.17: 7311470-7311470 on Build GRCh38

View Full Product Details