Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 29728529-29728790 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CABP7
Entrez Gene ID
164633
Gene Name
calcium binding protein 7
Gene Aliases
CALN2
UniGene
Hs.643608
Gene Chromosome Location
Chr.22: 29720090-29731833 on Build GRCh38
Transcripts
NM_182527.2 XM_006724161.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7221050 c.254-5A>T p.? Chr.22: 29728625-29728625 on Build GRCh38
6038535 c.286A>G p.T96A Chr.22: 29728662-29728662 on Build GRCh38
5455001 c.255T>C p.G85G Chr.22: 29728631-29728631 on Build GRCh38
3553202 c.256G>A p.D86N Chr.22: 29728632-29728632 on Build GRCh38
231935 c.296G>A p.G99E Chr.22: 29728672-29728672 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765267011 Chr.22: 29728770-29728770 on Build GRCh38
rs776840739 Chr.22: 29728663-29728663 on Build GRCh38
rs753150070 Chr.22: 29728579-29728579 on Build GRCh38
rs767699592 Chr.22: 29728747-29728747 on Build GRCh38
rs775268057 Chr.22: 29728626-29728626 on Build GRCh38
rs765824220 Chr.22: 29728671-29728671 on Build GRCh38
rs550357462 Chr.22: 29728568-29728568 on Build GRCh38
rs1004807311 Chr.22: 29728564-29728591 on Build GRCh38
rs199959463 Chr.22: 29728637-29728637 on Build GRCh38
rs762182207 Chr.22: 29728722-29728723 on Build GRCh38
rs753169975 Chr.22: 29728676-29728676 on Build GRCh38
rs140096 Chr.22: 29728625-29728625 on Build GRCh38
rs776700083 Chr.22: 29728762-29728762 on Build GRCh38
rs749239092 Chr.22: 29728729-29728729 on Build GRCh38
rs776525071 Chr.22: 29728593-29728593 on Build GRCh38
rs376748886 Chr.22: 29728747-29728747 on Build GRCh38
rs774567898 Chr.22: 29728745-29728745 on Build GRCh38
rs759508338 Chr.22: 29728665-29728665 on Build GRCh38
rs768600661 Chr.22: 29728630-29728630 on Build GRCh38
rs878901288 Chr.22: 29728662-29728662 on Build GRCh38
rs761953884 Chr.22: 29728749-29728749 on Build GRCh38
rs370905980 Chr.22: 29728609-29728609 on Build GRCh38
rs898046880 Chr.22: 29728577-29728577 on Build GRCh38
rs980498520 Chr.22: 29728696-29728696 on Build GRCh38
rs747948823 Chr.22: 29728758-29728758 on Build GRCh38
rs764448101 Chr.22: 29728702-29728702 on Build GRCh38
rs780676290 Chr.22: 29728715-29728715 on Build GRCh38
rs774342279 Chr.22: 29728746-29728746 on Build GRCh38
rs758742605 Chr.22: 29728583-29728583 on Build GRCh38
rs532674321 Chr.22: 29728718-29728718 on Build GRCh38
rs746696717 Chr.22: 29728608-29728608 on Build GRCh38
rs368548305 Chr.22: 29728769-29728769 on Build GRCh38
rs780766875 Chr.22: 29728620-29728620 on Build GRCh38
rs766692524 Chr.22: 29728644-29728644 on Build GRCh38
rs933262220 Chr.22: 29728721-29728721 on Build GRCh38
rs200634649 Chr.22: 29728719-29728719 on Build GRCh38
rs758863345 Chr.22: 29728697-29728697 on Build GRCh38
rs890628478 Chr.22: 29728555-29728555 on Build GRCh38
rs969490595 Chr.22: 29728681-29728681 on Build GRCh38
rs745357483 Chr.22: 29728624-29728624 on Build GRCh38
rs760988159 Chr.22: 29728643-29728643 on Build GRCh38
rs751864377 Chr.22: 29728711-29728711 on Build GRCh38
rs756962602 Chr.22: 29728714-29728714 on Build GRCh38

View Full Product Details