Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 44619022-44619265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD82
Entrez Gene ID
3732
Gene Name
CD82 molecule
Gene Aliases
4F9, C33, GR15, IA4, KAI1, R2, SAR2, ST6, TSPAN27
UniGene
Hs.527778
Gene Chromosome Location
Chr.11: 44565591-44620363 on Build GRCh38
Transcripts
NM_001024844.1 NM_002231.3 XM_006718223.2 XM_011520067.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
926999 c.727-1G>A p.? Chr.11: 44619048-44619048 on Build GRCh38
1979356 c.759C>T p.C253C Chr.11: 44619081-44619081 on Build GRCh38
6623907 c.765C>T p.H255H Chr.11: 44619087-44619087 on Build GRCh38
5619305 c.768C>G p.V256V Chr.11: 44619090-44619090 on Build GRCh38
5634261 c.750C>T p.I250I Chr.11: 44619072-44619072 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777044032 Chr.11: 44619145-44619145 on Build GRCh38
rs778859812 Chr.11: 44619177-44619177 on Build GRCh38
rs780577629 Chr.11: 44619127-44619127 on Build GRCh38
rs74885383 Chr.11: 44619066-44619066 on Build GRCh38
rs765960487 Chr.11: 44619156-44619156 on Build GRCh38
rs767273646 Chr.11: 44619160-44619160 on Build GRCh38
rs376795165 Chr.11: 44619138-44619138 on Build GRCh38
rs747327385 Chr.11: 44619132-44619132 on Build GRCh38
rs148874083 Chr.11: 44619117-44619117 on Build GRCh38
rs893249517 Chr.11: 44619242-44619242 on Build GRCh38
rs201405906 Chr.11: 44619082-44619082 on Build GRCh38
rs751291553 Chr.11: 44619105-44619105 on Build GRCh38
rs386753094 Chr.11: 44619242-44619243 on Build GRCh38
rs547161316 Chr.11: 44619191-44619191 on Build GRCh38
rs903202467 Chr.11: 44619187-44619187 on Build GRCh38
rs778967682 Chr.11: 44619070-44619070 on Build GRCh38
rs751381541 Chr.11: 44619157-44619157 on Build GRCh38
rs376175476 Chr.11: 44619151-44619151 on Build GRCh38
rs7107335 Chr.11: 44619090-44619090 on Build GRCh38
rs777837925 Chr.11: 44619057-44619057 on Build GRCh38
rs147912305 Chr.11: 44619115-44619115 on Build GRCh38
rs952097761 Chr.11: 44619181-44619181 on Build GRCh38
rs77781241 Chr.11: 44619243-44619243 on Build GRCh38
rs772351881 Chr.11: 44619126-44619126 on Build GRCh38
rs947715017 Chr.11: 44619233-44619233 on Build GRCh38
rs201671713 Chr.11: 44619116-44619116 on Build GRCh38
rs142351354 Chr.11: 44619088-44619088 on Build GRCh38
rs992402871 Chr.11: 44619225-44619225 on Build GRCh38
rs916792094 Chr.11: 44619230-44619230 on Build GRCh38
rs370925717 Chr.11: 44619147-44619147 on Build GRCh38
rs773237459 Chr.11: 44619122-44619122 on Build GRCh38
rs746076379 Chr.11: 44619081-44619081 on Build GRCh38
rs545998821 Chr.11: 44619083-44619083 on Build GRCh38
rs781061734 Chr.11: 44619112-44619112 on Build GRCh38
rs529985982 Chr.11: 44619214-44619214 on Build GRCh38
rs754925345 Chr.11: 44619106-44619106 on Build GRCh38
rs983588278 Chr.11: 44619192-44619192 on Build GRCh38
rs766230830 Chr.11: 44619097-44619097 on Build GRCh38
rs4755266 Chr.11: 44619242-44619242 on Build GRCh38
rs756070640 Chr.11: 44619170-44619170 on Build GRCh38
rs762518913 Chr.11: 44619152-44619152 on Build GRCh38
rs1043512248 Chr.11: 44619099-44619099 on Build GRCh38
rs754907833 Chr.11: 44619049-44619049 on Build GRCh38
rs1043378277 Chr.11: 44619237-44619237 on Build GRCh38
rs939626355 Chr.11: 44619224-44619224 on Build GRCh38
rs779803792 Chr.11: 44619040-44619040 on Build GRCh38
rs761266322 Chr.11: 44619087-44619087 on Build GRCh38
rs201650491 Chr.11: 44619096-44619096 on Build GRCh38
rs749272629 Chr.11: 44619064-44619064 on Build GRCh38
rs752630534 Chr.11: 44619163-44619163 on Build GRCh38
rs747008804 Chr.11: 44619041-44619041 on Build GRCh38

View Full Product Details