Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 64716658-64716931 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNS
Entrez Gene ID
2799
Gene Name
glucosamine (N-acetyl)-6-sulfatase
Gene Aliases
G6S
UniGene
Hs.334534
Gene Chromosome Location
Chr.12: 64713442-64759446 on Build GRCh38
Transcripts
NM_002076.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1363629 c.1618G>A p.G540S Chr.12: 64716782-64716782 on Build GRCh38
5554258 c.1589T>C p.F530S Chr.12: 64716811-64716811 on Build GRCh38
4470155 c.1643C>T p.S548F Chr.12: 64716757-64716757 on Build GRCh38
7266684 c.1598G>A p.R533H Chr.12: 64716802-64716802 on Build GRCh38
4044165 c.1647A>C p.K549N Chr.12: 64716753-64716753 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142001509 Chr.12: 64716704-64716704 on Build GRCh38
rs777360267 Chr.12: 64716751-64716751 on Build GRCh38
rs1041395672 Chr.12: 64716775-64716775 on Build GRCh38
rs202228620 Chr.12: 64716806-64716806 on Build GRCh38
rs941776434 Chr.12: 64716767-64716767 on Build GRCh38
rs746730219 Chr.12: 64716787-64716787 on Build GRCh38
rs1012454282 Chr.12: 64716816-64716816 on Build GRCh38
rs762354606 Chr.12: 64716686-64716686 on Build GRCh38
rs2279598 Chr.12: 64716903-64716903 on Build GRCh38
rs763486351 Chr.12: 64716839-64716839 on Build GRCh38
rs751302293 Chr.12: 64716840-64716841 on Build GRCh38
rs760916879 Chr.12: 64716716-64716716 on Build GRCh38
rs892889197 Chr.12: 64716770-64716770 on Build GRCh38
rs61743822 Chr.12: 64716802-64716802 on Build GRCh38
rs745869300 Chr.12: 64716705-64716705 on Build GRCh38
rs113381844 Chr.12: 64716740-64716740 on Build GRCh38
rs778448292 Chr.12: 64716771-64716771 on Build GRCh38
rs756752200 Chr.12: 64716766-64716766 on Build GRCh38
rs768509687 Chr.12: 64716790-64716790 on Build GRCh38
rs867942380 Chr.12: 64716864-64716864 on Build GRCh38
rs751913045 Chr.12: 64716746-64716746 on Build GRCh38
rs145017402 Chr.12: 64716739-64716739 on Build GRCh38
rs766745346 Chr.12: 64716742-64716742 on Build GRCh38
rs567918606 Chr.12: 64716893-64716893 on Build GRCh38
rs753506630 Chr.12: 64716759-64716759 on Build GRCh38
rs766615352 Chr.12: 64716784-64716784 on Build GRCh38
rs765412748 Chr.12: 64716731-64716731 on Build GRCh38
rs763232728 Chr.12: 64716773-64716773 on Build GRCh38
rs776925981 Chr.12: 64716791-64716791 on Build GRCh38
rs547985831 Chr.12: 64716889-64716889 on Build GRCh38
rs745781388 Chr.12: 64716779-64716779 on Build GRCh38
rs775194425 Chr.12: 64716713-64716713 on Build GRCh38
rs771883330 Chr.12: 64716706-64716706 on Build GRCh38
rs548014151 Chr.12: 64716892-64716892 on Build GRCh38
rs2230292 Chr.12: 64716750-64716750 on Build GRCh38
rs369482033 Chr.12: 64716730-64716730 on Build GRCh38
rs527899417 Chr.12: 64716795-64716795 on Build GRCh38
rs138355639 Chr.12: 64716817-64716817 on Build GRCh38
rs766104697 Chr.12: 64716737-64716738 on Build GRCh38
rs374071486 Chr.12: 64716883-64716883 on Build GRCh38
rs771940326 Chr.12: 64716782-64716782 on Build GRCh38
rs768718890 Chr.12: 64716721-64716721 on Build GRCh38

View Full Product Details