Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 114576591-114576852 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCAS2
Entrez Gene ID
10286
Gene Name
breast carcinoma amplified sequence 2
Gene Aliases
DAM1, SPF27, Snt309
UniGene
Hs.22960
Gene Chromosome Location
Chr.1: 114567557-114581644 on Build GRCh38
Transcripts
NM_005872.2

SNP Detail

SNP ID Chromosome Location
rs745698220 Chr.1: 114576705-114576705 on Build GRCh38
rs748943145 Chr.1: 114576720-114576720 on Build GRCh38
rs745847787 Chr.1: 114576790-114576790 on Build GRCh38
rs879157839 Chr.1: 114576643-114576643 on Build GRCh38
rs752903973 Chr.1: 114576772-114576772 on Build GRCh38
rs41274118 Chr.1: 114576771-114576771 on Build GRCh38
rs775083042 Chr.1: 114576719-114576719 on Build GRCh38
rs556776639 Chr.1: 114576761-114576761 on Build GRCh38
rs768093804 Chr.1: 114576722-114576722 on Build GRCh38
rs1036707399 Chr.1: 114576656-114576656 on Build GRCh38
rs990319570 Chr.1: 114576802-114576802 on Build GRCh38
rs779330744 Chr.1: 114576641-114576641 on Build GRCh38
rs376283933 Chr.1: 114576783-114576783 on Build GRCh38
rs773904549 Chr.1: 114576741-114576741 on Build GRCh38
rs746511965 Chr.1: 114576774-114576775 on Build GRCh38
rs754575699 Chr.1: 114576790-114576790 on Build GRCh38
rs779836704 Chr.1: 114576795-114576795 on Build GRCh38
rs779653387 Chr.1: 114576767-114576767 on Build GRCh38
rs573780560 Chr.1: 114576823-114576823 on Build GRCh38
rs770834209 Chr.1: 114576688-114576688 on Build GRCh38
rs1051726286 Chr.1: 114576821-114576821 on Build GRCh38
rs369965672 Chr.1: 114576765-114576765 on Build GRCh38
rs1048922410 Chr.1: 114576714-114576714 on Build GRCh38
rs769652213 Chr.1: 114576713-114576713 on Build GRCh38
rs752995608 Chr.1: 114576645-114576645 on Build GRCh38
rs768359627 Chr.1: 114576809-114576809 on Build GRCh38
rs764256314 Chr.1: 114576776-114576776 on Build GRCh38
rs780941728 Chr.1: 114576788-114576788 on Build GRCh38
rs755840385 Chr.1: 114576791-114576791 on Build GRCh38
rs946308648 Chr.1: 114576797-114576797 on Build GRCh38
rs201857003 Chr.1: 114576763-114576763 on Build GRCh38
rs781038404 Chr.1: 114576689-114576689 on Build GRCh38
rs746906940 Chr.1: 114576678-114576678 on Build GRCh38
rs758515230 Chr.1: 114576672-114576672 on Build GRCh38
rs150004706 Chr.1: 114576764-114576764 on Build GRCh38
rs538053862 Chr.1: 114576711-114576711 on Build GRCh38
rs557452123 Chr.1: 114576806-114576806 on Build GRCh38
rs377453007 Chr.1: 114576676-114576676 on Build GRCh38
rs1028441213 Chr.1: 114576808-114576808 on Build GRCh38
rs761236968 Chr.1: 114576750-114576750 on Build GRCh38
rs990420305 Chr.1: 114576775-114576775 on Build GRCh38
rs750467066 Chr.1: 114576755-114576755 on Build GRCh38
rs751658982 Chr.1: 114576780-114576780 on Build GRCh38

View Full Product Details