Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 161510379-161510614 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSTL5
Entrez Gene ID
56884
Gene Name
follistatin like 5
Gene Aliases
-
UniGene
Hs.32452
Gene Chromosome Location
Chr.4: 161383892-162164034 on Build GRCh38
Transcripts
NM_001128427.2 NM_001128428.2 NM_020116.4 XM_011532126.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4763291 c.1315G>T p.A439S Chr.4: 161510422-161510422 on Build GRCh38
5650936 c.1339+2T>C p.? Chr.4: 161510396-161510396 on Build GRCh38
4927594 c.1336G>T p.E446* Chr.4: 161510401-161510401 on Build GRCh38
281260 c.1333G>T p.E445* Chr.4: 161510404-161510404 on Build GRCh38
4847404 c.1336G>A p.E446K Chr.4: 161510401-161510401 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764409024 Chr.4: 161510449-161510449 on Build GRCh38
rs761075001 Chr.4: 161510435-161510435 on Build GRCh38
rs769702826 Chr.4: 161510413-161510413 on Build GRCh38
rs574233482 Chr.4: 161510571-161510575 on Build GRCh38
rs72683746 Chr.4: 161510543-161510543 on Build GRCh38
rs772458745 Chr.4: 161510571-161510571 on Build GRCh38
rs938126961 Chr.4: 161510486-161510486 on Build GRCh38
rs775974271 Chr.4: 161510434-161510434 on Build GRCh38
rs945610342 Chr.4: 161510466-161510466 on Build GRCh38
rs781394124 Chr.4: 161510456-161510457 on Build GRCh38
rs896497461 Chr.4: 161510528-161510528 on Build GRCh38
rs143070893 Chr.4: 161510472-161510472 on Build GRCh38
rs748944406 Chr.4: 161510425-161510425 on Build GRCh38
rs1026992349 Chr.4: 161510555-161510555 on Build GRCh38
rs1056372653 Chr.4: 161510519-161510519 on Build GRCh38
rs78459880 Chr.4: 161510475-161510475 on Build GRCh38
rs10019848 Chr.4: 161510465-161510465 on Build GRCh38
rs748020251 Chr.4: 161510411-161510411 on Build GRCh38
rs550627823 Chr.4: 161510433-161510433 on Build GRCh38
rs201799022 Chr.4: 161510432-161510432 on Build GRCh38
rs748221322 Chr.4: 161510467-161510468 on Build GRCh38
rs754111389 Chr.4: 161510462-161510462 on Build GRCh38
rs532068920 Chr.4: 161510509-161510509 on Build GRCh38
rs905283231 Chr.4: 161510481-161510481 on Build GRCh38
rs372450375 Chr.4: 161510427-161510427 on Build GRCh38
rs770454103 Chr.4: 161510426-161510426 on Build GRCh38
rs188964014 Chr.4: 161510491-161510491 on Build GRCh38
rs755132027 Chr.4: 161510439-161510447 on Build GRCh38
rs1013716103 Chr.4: 161510567-161510567 on Build GRCh38
rs773050251 Chr.4: 161510424-161510424 on Build GRCh38

View Full Product Details