Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 11353713-11353958 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC159
Entrez Gene ID
126075
Gene Name
coiled-coil domain containing 159
Gene Aliases
-
UniGene
Hs.631636
Gene Chromosome Location
Chr.19: 11346340-11354951 on Build GRCh38
Transcripts
NM_001080503.2 XM_006722643.1 XM_011527678.2 XM_017026255.1 XM_017026256.1 XM_017026257.1 XM_017026258.1 XM_017026259.1 XM_017026260.1 XM_017026261.1 XM_017026262.1 XM_017026263.1 XM_017026264.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4832289 c.692C>G p.S231C Chr.19: 11353794-11353794 on Build GRCh38
4832290 c.1037C>G p.S346C Chr.19: 11353794-11353794 on Build GRCh38
1390330 c.1056G>A p.Q352Q Chr.19: 11353813-11353813 on Build GRCh38
2814281 c.1071C>T p.S357S Chr.19: 11353828-11353828 on Build GRCh38
1390329 c.711G>A p.Q237Q Chr.19: 11353813-11353813 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748284386 Chr.19: 11353844-11353844 on Build GRCh38
rs770907137 Chr.19: 11353864-11353864 on Build GRCh38
rs935552738 Chr.19: 11353795-11353795 on Build GRCh38
rs774205788 Chr.19: 11353788-11353788 on Build GRCh38
rs749499281 Chr.19: 11353856-11353856 on Build GRCh38
rs1026254354 Chr.19: 11353772-11353772 on Build GRCh38
rs776772499 Chr.19: 11353746-11353746 on Build GRCh38
rs762874909 Chr.19: 11353779-11353779 on Build GRCh38
rs963401109 Chr.19: 11353738-11353738 on Build GRCh38
rs577539469 Chr.19: 11353914-11353914 on Build GRCh38
rs769955664 Chr.19: 11353845-11353845 on Build GRCh38
rs377572115 Chr.19: 11353804-11353804 on Build GRCh38
rs752060160 Chr.19: 11353833-11353833 on Build GRCh38
rs777772554 Chr.19: 11353900-11353900 on Build GRCh38
rs377320714 Chr.19: 11353825-11353825 on Build GRCh38
rs755518502 Chr.19: 11353842-11353842 on Build GRCh38
rs375449767 Chr.19: 11353863-11353863 on Build GRCh38
rs748113650 Chr.19: 11353762-11353762 on Build GRCh38
rs770820843 Chr.19: 11353782-11353782 on Build GRCh38
rs200561735 Chr.19: 11353821-11353821 on Build GRCh38
rs371059805 Chr.19: 11353805-11353805 on Build GRCh38
rs778182058 Chr.19: 11353847-11353847 on Build GRCh38
rs959570844 Chr.19: 11353837-11353837 on Build GRCh38
rs369884820 Chr.19: 11353766-11353766 on Build GRCh38
rs759504170 Chr.19: 11353794-11353794 on Build GRCh38
rs775239404 Chr.19: 11353901-11353901 on Build GRCh38
rs568436297 Chr.19: 11353765-11353765 on Build GRCh38
rs780370816 Chr.19: 11353830-11353830 on Build GRCh38
rs774191139 Chr.19: 11353867-11353867 on Build GRCh38
rs781497063 Chr.19: 11353843-11353843 on Build GRCh38
rs759185256 Chr.19: 11353890-11353890 on Build GRCh38
rs535780699 Chr.19: 11353893-11353893 on Build GRCh38
rs373080827 Chr.19: 11353801-11353801 on Build GRCh38
rs569832224 Chr.19: 11353898-11353899 on Build GRCh38
rs982276637 Chr.19: 11353751-11353751 on Build GRCh38

View Full Product Details