Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48791085-48791358 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATA1
Entrez Gene ID
2623
Gene Name
GATA binding protein 1
Gene Aliases
ERYF1, GATA-1, GF-1, GF1, NF-E1, NFE1, XLANP, XLTDA, XLTT
UniGene
Hs.765
Gene Chromosome Location
Chr.X: 48786540-48794311 on Build GRCh38
Transcripts
NM_002049.3 XM_011543897.2 XM_011543898.2

SNP Detail

SNP ID Chromosome Location
rs782299679 Chr.X: 48791282-48791282 on Build GRCh38
rs201489369 Chr.X: 48791258-48791258 on Build GRCh38
rs782591058 Chr.X: 48791271-48791271 on Build GRCh38
rs782155672 Chr.X: 48791216-48791216 on Build GRCh38
rs142614402 Chr.X: 48791267-48791267 on Build GRCh38
rs139614533 Chr.X: 48791283-48791283 on Build GRCh38
rs782698349 Chr.X: 48791203-48791203 on Build GRCh38
rs398124628 Chr.X: 48791282-48791283 on Build GRCh38
rs587776451 Chr.X: 48791111-48791111 on Build GRCh38
rs149753411 Chr.X: 48791305-48791305 on Build GRCh38
rs892837818 Chr.X: 48791226-48791226 on Build GRCh38
rs781856701 Chr.X: 48791254-48791254 on Build GRCh38
rs782366452 Chr.X: 48791273-48791273 on Build GRCh38
rs782214998 Chr.X: 48791128-48791128 on Build GRCh38
rs150572851 Chr.X: 48791272-48791272 on Build GRCh38
rs587776452 Chr.X: 48791329-48791329 on Build GRCh38
rs374300356 Chr.X: 48791321-48791321 on Build GRCh38
rs782697197 Chr.X: 48791219-48791219 on Build GRCh38
rs782408441 Chr.X: 48791145-48791145 on Build GRCh38
rs61753429 Chr.X: 48791310-48791310 on Build GRCh38
rs782411199 Chr.X: 48791294-48791294 on Build GRCh38
rs1045976499 Chr.X: 48791157-48791157 on Build GRCh38
rs398124627 Chr.X: 48791296-48791297 on Build GRCh38
rs782341488 Chr.X: 48791306-48791306 on Build GRCh38
rs139200954 Chr.X: 48791174-48791174 on Build GRCh38
rs781808940 Chr.X: 48791327-48791327 on Build GRCh38
rs782188059 Chr.X: 48791173-48791173 on Build GRCh38
rs782691816 Chr.X: 48791339-48791339 on Build GRCh38
rs1029589980 Chr.X: 48791318-48791318 on Build GRCh38
rs782165033 Chr.X: 48791337-48791337 on Build GRCh38
rs141512330 Chr.X: 48791319-48791319 on Build GRCh38
rs782258887 Chr.X: 48791138-48791138 on Build GRCh38
rs781957095 Chr.X: 48791186-48791186 on Build GRCh38
rs12841023 Chr.X: 48791160-48791160 on Build GRCh38
rs782678694 Chr.X: 48791259-48791259 on Build GRCh38
rs782744233 Chr.X: 48791225-48791225 on Build GRCh38
rs781912832 Chr.X: 48791301-48791301 on Build GRCh38
rs782038466 Chr.X: 48791149-48791149 on Build GRCh38
rs782474159 Chr.X: 48791224-48791224 on Build GRCh38
rs782294292 Chr.X: 48791263-48791263 on Build GRCh38
rs372131208 Chr.X: 48791222-48791222 on Build GRCh38
rs587776453 Chr.X: 48791330-48791330 on Build GRCh38
rs782628386 Chr.X: 48791134-48791134 on Build GRCh38

View Full Product Details